Skip to Content
Merck
All Photos(4)

Key Documents

SAB4200080

Sigma-Aldrich

Anti-Vinculin Antibody

mouse monoclonal, V284

Synonym(s):

Anti-CMD1W, Anti-MVCL, Anti-VCL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Vinculin antibody, Mouse monoclonal, clone V284, purified from hybridoma cell culture

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture
purified immunoglobulin

antibody product type

primary antibodies

clone

V284, monoclonal

form

buffered aqueous solution

mol wt

antigen ~117 kDa

species reactivity

rabbit, chicken, human, rat, monkey, mouse, canine

concentration

~1 mg/mL

technique(s)

immunocytochemistry: 4-8 μg/mL using HeLa cells fixed with methanol:acetone
immunoprecipitation (IP): suitable
western blot: 0.005-0.01 μg/mL using HeLa total cell extracts

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VCL(7414)
mouse ... Vcl(22330)
rat ... Vcl(305679)

General description

Monoclonal Anti-Vinculin (mouse IgG1 isotype) is derived from the hybridoma V284 produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with purified human platelet vinculin. In muscle, vinculin is localized in the fascia adherens of the intercalated disk (cardiac muscle), neuromuscular junctions, myotendinous junctions (skeletal muscle) and the membrane-associated dense bodies of smooth muscle.
Vinculin (VCL) is a ubiquitously expressed cytoskeletal protein, encoded by the gene mapped to human chromosome 10q11.2-qter. VCL is a vital element of adherens type cell junctions. Vinculin structure contains a globular head connected to a tail domain by a short proline-rich sequence, and an intramolecular interaction between the head and tail hides the various ligand-binding sites in the protein.

Immunogen

purified human platelet vinculin.

Application

Anti-Vinculin antibody, Mouse monoclonal has been used:
  • to probe blots in immunoblotting
  • immunochemical staining
  • immunoprecipitation
  • immunocytochemistry

Monoclonal Anti-Vinculin antibody produced in mouse has been used in western blotting.

Biochem/physiol Actions

Vinculin (VCL) is a cytoskeletal protein that supports cell-to-cell and cell to matrix junctions. It is also essentially involved in normal embryonic development and cardiac function, due to its role in controlling adhesion and motility, as well as cell spreading. In mature focal adhesions (FAs), VCL acts as an important constituent of the “molecular clutch” that induces the transmission of force from cytoplasmic F-actin to membrane-bound integrins. Aberration or mutation of VCL leads to cardiomyopathy, which is associated with hypertension. experimental studies reveal that, VCL might be used as a biomarker of aging or heart failure. Vinculin associates with FAs and assists in several cellular functions, such as stabilizing and strengthening FAs and promoting their maturation, extracellular matrix (ECM) mechanosensing, regulating actin cytoskeletal dynamics and signaling pathways involved in apoptosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

STAT3 localizes to the ER, acting as a gatekeeper for ER-mitochondrion Ca2+ fluxes and apoptotic responses.
Avalle L, et al.
Cell Death and Differentiation, 26(5), 932-942 (2019)
The mechanical and biological properties of polycarbonate-modified F127 hydrogels after incorporating active pendent double-bonds
Wang T, et al.
Polymer, 57, 21-28 (2015)
Sex-specific association of rs4746172 of VCL gene with hypertension in two Han populations from Southern China.
Yu Q
Scientific Reports (2015)
In-Depth, Label-Free Analysis of the Erythrocyte Cytoplasmic Proteome in Diamond Blackfan Anemia Identifies a Unique Inflammatory Signature
Pesciotta EN
PLoS ONE, 10 (2015)
Increased formate overflow is a hallmark of oxidative cancer
Meiser J, et al.
Nature Communications, 9(1), 1368-1368 (2018)

Articles

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Quantitative and qualitative western blotting to validate knockdown by esiRNA.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service