Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500954

Sigma-Aldrich

Anti-ZIMP7 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
polyclonal
application:
ELISA
species reactivity:
human, mouse
technique(s):
ELISA: suitable
immunoblotting: suitable
citations:
4

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ZMIZ2(83637)

Immunogen

Antibody was raised against a 17 amino acid synthetic peptide near the center of human ZIMP7.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500954.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason Beliakoff et al.
Nuclear receptor signaling, 4, e017-e017 (2006-07-25)
The androgen receptor (AR) plays a critical role in male sexual development and in normal and malignant prostate cell growth and survival. It has been shown that AR transcriptional activation is regulated through interactions with a variety of transcriptional co-regulators.
N Ueki et al.
Nature biotechnology, 16(13), 1338-1342 (1998-12-16)
Nuclear proteins have essential roles in cell proliferation and differentiation. We have developed a yeast selection system-the nuclear transportation trap (NTT)-to identify genes encoding nuclear transport signals. Both unknown and previously identified nuclear localization signals were identified from a human
Yue Peng et al.
The Journal of biological chemistry, 285(15), 11465-11475 (2010-02-18)
The PIAS proteins (protein inhibitor of activated STAT) were originally identified as inhibitors of the JAK-STAT pathway. Subsequently, their roles on transcriptional regulation have been identified in modulation of the androgen receptor (AR) and other nuclear hormone receptor-mediated actions. Zimp7
Chun-Yin Huang et al.
Molecular endocrinology (Baltimore, Md.), 19(12), 2915-2929 (2005-07-30)
Members of the PIAS (protein inhibitor of activated signal transducer and activator of transcription) family are negative regulators of the Janus family of tyrosine kinase (JAK)-signal transducer and activator of transcription pathway. Recently, PIAS proteins have been shown to interact

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service