Skip to Content
Merck
All Photos(3)

Key Documents

SAB2500328

Sigma-Aldrich

Anti-DPP4/CD26 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ADABP, Anti-ADCP2, Anti-Adenosine deaminase complexing protein 2, Anti-CD26, Anti-Dipeptidyl-peptidase 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat, canine

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DPP4(1803)

General description

Dipeptidyl peptidase 4 (DPP4), also known as cluster of differentiation 26 (CD26), is localized on T- cell surface and interacts with adenosine deaminase (ADA), collagen and CD459. The gene encoding this protein is localized on human chromosome 2q24.2.

Immunogen

Peptide with sequence C-PPHFDKSKKYP from the internal region of the protein sequence according to NP_001926.2.

Biochem/physiol Actions

Dipeptidyl peptidase 4 (DPP4) or CD26 plays a vital role in T cell co-stimulation, cell-to-cell adhesion and in human immunodeficiency virus (HIV) infection. It functions as a serine protease enzyme that catalyzes the cleavage of dipeptides from peptides containing proline or alanine residues. This protein is a biomarker and therapeutic target for asthma. Deletion of the DPP4 gene affects carbohydrate metabolism which in turn leads to mental retardation and hypotonia. Abnormal expression of the protein is observed during liver regeneration, cirrhosis and liver tumorigenesis. Also, DPP4 level is reduced in serum of chronic obstructive pulmonary disease (COPD) patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natacha Bohin et al.
Stem cell reports, 15(1), 156-170 (2020-06-13)
Intestinal crypts have great capacity for repair and regeneration after intestinal stem cell (ISC) injury. Here, we define the cellular remodeling process resulting from ISC niche interruption by transient Notch pathway inhibition in adult mice. Although ISCs were retained, lineage
A novel deletion in 2q24.1q24.2 in a girl with mental retardation and generalized hypotonia: a case report.
Palumbo O
Molecular Cytogenetics (2012)
Molecular analyses of human and rat dipeptidyl peptidase IV.
Abbott CA
Advances in Experimental Medicine and Biology (1997)
Dipeptidyl peptidase-4 is highly expressed in bronchial epithelial cells of untreated asthma and it increases cell proliferation along with fibronectin production in airway constitutive cells.
Shiobara T
Respiratory Research (2016)
Genomic organization, exact localization, and tissue expression of the human CD26 (dipeptidyl peptidase IV) gene.
Abbott CA
Immunogenetics (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service