Skip to Content
Merck
All Photos(2)

Key Documents

SAB2108557

Sigma-Aldrich

Anti-LOR

IgG fraction of antiserum

Synonym(s):

Anti-MGC111513

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

35 kDa

species reactivity

human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_000427

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LOR(4014)

General description

The LOR gene codes for a a small basic protein, loricrin. It is predominantly localized to the cornified cell envelope.

Immunogen

Synthetic peptide directed towards the N terminal region of human LOR

Biochem/physiol Actions

LOR is a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in this gene are associated with Vohwinkel′s syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.LOR encodes loricrin, a major protein component of the cornified cell envelope found in terminally differentiated epidermal cells. Mutations in LOR may be the cause of both Vohwinkel′s syndrome and progressive symmetric erythrokeratoderma, both inherited skin diseases.
Mutations in the gene LOR, leads to an autosomal dominant genodermatosis called loricrin keratoderma. The disease is characterized by palmoplantar keratoderma and ichthyosis. Variation in the gene also results in Vohwinkel syndrome with ichthyosis and progressive symmetrical erythrokeratodermia, an inherited cornification disorder.

Sequence

Synthetic peptide located within the following region: GYSGGGCGGGSSGGGGGGGIGGCGGGSGGSVKYSGGGGSSGGGSGCFSSG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel autosomal dominant mutation in loricrin presenting as prominent ichthyosis.
Pohler E, et al.
British Journal of Dermatology, 173(5), 1291-1294 (2015)
Evidence for the absence of mutations at GJB3, GJB4 and LOR in progressive symmetrical erythrokeratodermia.
Wei S, et al.
Clinical and Experimental Dermatology, 36(4), 399-405 (2011)
Activation of vascular endothelial growth factor receptor 2 in a cellular model of loricrin keratoderma.
Yoneda K, et al.
The Journal of Biological Chemistry (2010)
Kinga Skieresz-Szewczyk et al.
Cells, 11(12) (2022-06-25)
The lingual mucosa in birds is covered with two specific types of multilayered epithelia, i.e., the para- and orthokeratinized epithelium, that differ structurally and functionally. Comprehensive information on proteins synthesized in keratinocyte during their cytodifferentiation in subsequent layers of multilayered

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service