Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100357

Sigma-Aldrich

Anti-CCK antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-Cholecystokinin, Anti-MGC117187

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

13 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCK(885)

General description

CCK codes for cholecystokinin. It is a brain/gut peptide. This gene is located on human chromosome 3p22.

Immunogen

Synthetic peptide directed towards the middle region of human CCK

Application

Anti-CCK antibody produced in rabbit has been used in staining.

Biochem/physiol Actions

Cholecystokinin (CCK) is a brain/gut peptide. In the gut, it induces the release of pancreatic enzymes and the contraction of the gallbladder. In the brain, its physiologic role is unclear. The cholecystokinin pro-hormone is processed by endo- and exo-proteolytic cleavages.

Sequence

Synthetic peptide located within the following region: IQQARKAPSGRMSIVKNLQNLDPSHRISDRDYMGWMDFGRRSAEEYEYPS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alteration of Interneuron Immunoreactivity and Autophagic Activity in Rat Hippocampus after Single High-Dose Whole-Brain Irradiation
Ouyang YB, et al.
Cureus, 9(6) (2017)
3p22. 1p21. 31 microdeletion identifies CCK as Asperger syndrome candidate gene and shows the way for therapeutic strategies in chromosome imbalances
Iourov IY, et al.
Molecular Cytogenetics, 8(1), 82-82 (2015)
Role of CCK/gastrin receptors in gastrointestinal/metabolic diseases and results of human studies using gastrin/CCK receptor agonists/antagonists in these diseases
Berna MJ, et al.
Current Topics in Medicinal Chemistry, 7(12), 1211-1211 (2007)
Cleavage of arginyl-arginine and lysyl-arginine from the C-terminus of pro-hormone peptides by human germinal angiotensin I-converting enzyme (ACE) and the C-domain of human somatic ACE
Isaac RE, et al.
The Biochemical Journal, 328(Pt), 2-2 (1997)
Mechanism of action of cholecystokinin: a not atypical brain-gut peptide
Williams JA
Nihon Naibunpi Gakkai Zasshi, 61(5), 533-540 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service