Skip to Content
Merck
All Photos(3)

Key Documents

SAB1408428

Sigma-Aldrich

Anti-MLKL antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ34389

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~54.5 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLKL(197259)

General description

MLKL has a C-terminal pseudokinase (PsK) domain and an N-terminal 4-helix bundle (4HB) domain. Both these domains are held together by two brace helices.
The gene MLKL (mixed lineage kinase domain-like protein) is mapped to human chromosome 16q23. Activated MLKL is present at the cell membrane.

Immunogen

MLKL (NP_689862.1, 1 a.a. ~ 471 a.a) full-length human protein.

Sequence
MENLKHIITLGQVIHKRCEEMKYCKKQCRRLGHRVLGLIKPLEMLQDQGKRSVPSEKLTTAMNRFKAALEEANGEIEKFSNRSNICRFLTASQDKILFKDVNRKLSDVWKELSLLLQVEQRMPVSPISQGASWAQEDQQDADEDRRAFQMLRRDNEKIEASLRRLEINMKEIKETLRQYLPPKCMQEIPQEQIKEIKKEQLSGSPWILLRENEVSTLYKGEYHRAPVAIKVFKKLQAGSIAIVRQTFNKEIKTMKKFESPNILRIFGICIDETVTPPQFSIVMEYCELGTLRELLDREKDLTLGKRMVLVLGAARGLYRLHHSEAPELHGKIRSSNFLVTQGYQVKLAGFELRKTQTSMSLGTTREKTDRVKSTAYLSPQELEDVFYQYDVKSEIYSFGIVLWEIATGDIPFQGCNSEKIRKLVAVKRQQEPLGEDCPSELREIIDECRAHDPSVRPSVDEILKKLSTFSK

Application

Anti-MLKL antibody produced in mouse has been used in western blotting.

Biochem/physiol Actions

MLKL (mixed lineage kinase domain-like protein) primarily causes receptor-interacting protein (RIP) kinase–dependent necroptosis. However, during hepatitis, it results in programmed hepatocellular necrosis which is independent of RIPK3. MLKL also participates in endosomal trafficking and the formation of extracellular vesicle.
MLKL activation and subsequent N-terminal 4HB domain unleashing leads to oligomerization and cell membrane entry. This in turn leads to necroptosis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nupur Bansal et al.
Scientific reports, 9(1), 16853-16853 (2019-11-16)
Mixed Lineage Kinase domain-Like (MLKL), a key player in necroptosis, is a multi-domain protein with an N-terminal 4 helical bundle (4HB) and a pseudokinase domain (PsK) connected by brace helices. Phosphorylation of PsK domain of MLKL is a key step
Depletion of RIPK3 or MLKL blocks TNF-driven necroptosis and switches towards a delayed RIPK1 kinase-dependent apoptosis.
Remijsen Q, et. al.
Cell Death & Disease, 5, e1004-e1004 (2014)
MLKL Activation Triggers NLRP3-Mediated Processing and Release of IL-1? Independently of Gasdermin-D.
Gutierrez KD
Journal of Immunology, 198, 2156-2156 (2017)
Kevin Puertas-Neyra et al.
Frontiers in neuroanatomy, 16, 812487-812487 (2022-03-01)
Retinal neurodegenerative diseases are the leading causes of visual impairment and irreversible blindness worldwide. Although the retinal response to injury remains closely similar between different retinal neurodegenerative diseases, available therapeutic alternatives are only palliative, too expensive, or very specific, such
MLKL, the Protein that Mediates Necroptosis, Also Regulates Endosomal Trafficking and Extracellular Vesicle Generation.
Yoon S
Immunity, 47, 51-51 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service