Skip to Content
Merck
All Photos(1)

Key Documents

SAB1407409

Sigma-Aldrich

Anti-CD209 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDSIGN, CLEC4L, DC-SIGN, DC-SIGN1, MGC129965

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~45.8 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD209(30835)

General description

Cluster of differentiation 209 (CD209), also known as dendritic cell-specific intercellular adhesion molecule-3-grabbing non-integrin (DC-SIGN), is part of the innate immune system. It is expressed by intestinal dendritic cells and alveolar macrophages. The gene encoding this C-type lectin contains seven exons and is localized on human chromosome 19p13.2.
This gene encodes a transmembrane receptor and is often referred to as DC-SIGN because of its expression on the surface of dendritic cells and macrophages. The encoded protein is involved in the innate immune system and recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses with a large impact on public health. The protein is organized into three distinct domains: an N-terminal transmembrane domain, a tandem-repeat neck domain and C-type lectin carbohydrate recognition domain. The extracellular region consisting of the C-type lectin and neck domains has a dual function as a pathogen recognition receptor and a cell adhesion receptor by binding carbohydrate ligands on the surface of microbes and endogenous cells. The neck region is important for homo-oligomerization which allows the receptor to bind multivalent ligands with high avidity. Variations in the number of 23 amino acid repeats in the neck domain of this protein are rare but have a significant impact on ligand binding ability. This gene is closely related in terms of both sequence and function to a neighboring gene (GeneID 10332; often referred to as L-SIGN). DC-SIGN and L-SIGN differ in their ligand-binding properties and distribution. Alternative splicing results in multiple variants.

Immunogen

CD209 (NP_066978.1, 1 a.a. ~ 404 a.a) full-length human protein.

Sequence
MSDSKEPRLQQLGLLEEEQLRGLGFRQTRGYKSLAGCLGHGPLVLQLLSFTLLAGLLVQVSKVPSSISQEQSRQDAIYQNLTQLKAAVGELSEKSKLQEIYQELTQLKAAVGELPEKSKLQEIYQELTRLKAAVGELPEKSKLQEIYQELTWLKAAVGELPEKSKMQEIYQELTRLKAAVGELPEKSKQQEIYQELTRLKAAVGELPEKSKQQEIYQELTRLKAAVGELPEKSKQQEIYQELTQLKAAVERLCHPCPWEWTFFQGNCYFMSNSQRNWHDSITACKEVGAQLVVIKSAEEQNFLQLQSSRSNRFTWMGLSDLNQEGTWQWVDGSPLLPSFKQYWNRGEPNNVGEEDCAEFSGNGWNDDKCNLAKFWICKKSAASCSRDEEQFLSPAPATPNPPPA

Biochem/physiol Actions

Cluster of differentiation 209 (CD209) mediates viral infection and activates immune responses. It activates T-cells and aids in their growth. The protein recognizes various pathogens and triggers immunosuppressive reactions. It is involved in the innate immune system. It recognizes numerous evolutionarily divergent pathogens ranging from parasites to viruses that have severe health impacts on humans. CD209/ DC-SIGN mediates the entry of various viruses such as dengue virus, human immunodeficiency virus (HIV), Ebola virus and human cytomegalovirus into the human cells. CD209 acts as an alternative receptor for SARS-CoV-2, a causative agent for the novel coronavirus disease 2019 (COVID-19).

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The activation of B cells enhances DC-SIGN expression and promotes susceptibility of B cells to HPAI H5N1 infection.
Na-Ek P, et al.
Biochemical and Biophysical Research Communications, 490(4), 1301-1306 (2017)
The association between CD209 gene polymorphisms and pulmonary tuberculosis susceptibility: a meta-analysis.
Yi L, et al.
International Journal of Clinical and Experimental Pathology, 8(10), 12437-12437 (2015)
Edit I Buzás et al.
Autoimmunity, 39(8), 691-704 (2006-12-21)
The immune system is a complex functional network of diverse cells and soluble molecules orchestrating innate and adaptive immunity. Biological information, to run these intricate interactions, is not only stored in protein sequences but also in the structure of the
Boonrat Tassaneetrithep et al.
The Journal of experimental medicine, 197(7), 823-829 (2003-04-12)
Dengue virus is a single-stranded, enveloped RNA virus that productively infects human dendritic cells (DCs) primarily at the immature stage of their differentiation. We now find that all four serotypes of dengue use DC-SIGN (CD209), a C-type lectin, to infect
Human DC-SIGN binds specific human milk glycans.
Noll AJ, et al.
The Biochemical Journal (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service