Skip to Content
Merck
All Photos(4)

Key Documents

SAB1405562

Sigma-Aldrich

Anti-CAT antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

MGC138422, MGC138424

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~59.8 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAT(847)

General description

Catalase (CAT) is an antioxidant enzyme. CAT gene is located on human chromosome 11p13. CAT is present in peroxisomes of most aerobic organisms. It is a heme enzyme expressed mainly in liver.

Immunogen

CAT (NP_001743.1, 1 a.a. ~ 527 a.a) full-length human protein.

Sequence
MADSRDPASDQMQHWKEQRAAQKADVLTTGAGNPVGDKLNVITVGPRGPLLVQDVVFTDEMAHFDRERIPERVVHAKGAGAFGYFEVTHDITKYSKAKVFEHIGKKTPIAVRFSTVAGESGSADTVRDPRGFAVKFYTEDGNWDLVGNNTPIFFIRDPILFPSFIHSQKRNPQTHLKDPDMVWDFWSLRPESLHQVSFLFSDRGIPDGHRHMNGYGSHTFKLVNANGEAVYCKFHYKTDQGIKNLSVEDAARLSQEDPDYGIRDLFNAIATGKYPSWTFYIQVMTFNQAETFPFNPFDLTKVWPHKDYPLIPVGKLVLNRNPVNYFAEVEQIAFDPSNMPPGIEASPDKMLQGRLFAYPDTHRHRLGPNYLHIPVNCPYRARVANYQRDGPMCMQDNQGGAPNYYPNSFGAPEQQPSALEHSIQYSGEVRRFNTANDDNVTQVRAFYVNVLNEEQRKRLCENIAGHLKDAQIFIQKKAVKNFTEVHPDYGSHIQALLDKYNAEKPKNAIHTFVQSGSHLAAREKANL

Biochem/physiol Actions

Catalase (CAT) protects different cell types from the harmful effects of hydrogen peroxide. It is involved in idiopathic male infertility. CAT converts hydrogen peroxide to water and molecular oxygen. CAT promotes resistance against many diseases. Mutations in CAT is linked to acatalasemia.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of catalase expression in healthy and cancerous cells
Glorieux C, et al.
Free Radical Biology & Medicine, 87(3) (2015)
A novel catalase mutation (a GA insertion) causes the Hungarian type of acatalasemia
Goth L , et al.
Blood Cells, Molecules and Diseases, 26(2) (2000)
A new type 1 diabetes susceptibility locus containing the catalase gene (chromosome 11p13) in a Russian population
Chistiakov D, et al.
Diabetes/metabolism Research and Reviews, 20(3) (2004)
The role of catalase C262T gene polymorphism in the susceptibility and survival of cancers
Wang C, et al.
Scientific Reports, 6(5), 26973-26973 (2016)
Structure and heme environment of beef liver catalase at 2.5 A resolution
Reid T, et al.
Proceedings of the National Academy of Sciences of the USA, 78(8) (1981)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service