Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402920

Sigma-Aldrich

Monoclonal Anti-SLC5A3 antibody produced in mouse

clone 3A6, purified immunoglobulin, buffered aqueous solution

Synonym(s):

SMIT, SMIT2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3A6, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.1 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC5A3(6526)

General description

Solute carrier family 5 member 3 (SLC5A3) is a member of SLC5A gene family, which shares five transmembrane segment inverted repeats of the LeuT structural family. SLC5A3 is expressed in brain, kidney and placenta. SLC5A3 gene is located on human chromosome 21q22.11.

Immunogen

SLC5A3 (NP_008864, 533 a.a. ~ 641 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
TPPPTKEQIRTTTFWSKKNLVVKENCSPKEEPYKMQEKSILRCSENNETINHIIPNGKSEDSIKGLQPEDVNLLVTCREEGNPVASLGHSEAETPVDAYSNGQAALMGE

Application

Monoclonal Anti-SLC5A3 antibody produced in mouse has been used in dSTORM (direct stochastic optical reconstruction microscopy).

Biochem/physiol Actions

Solute carrier family 5 member 3 (SLC5A3) functions in cellular osmoregulation. Overexpression of SLC5A3 contributes to pathophysiology of Down syndrome. SLC5A3 acts as a transporter in hypotonic volume regulation of mammalian cells. SLC5A3 regulates millimolar intracellular concentrations of myo-inositol and promotes transepithelial myo-inositol transport in kidney, intestine, retina and choroid plexus.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The structural organization of the human Na+/myo-inositol cotransporter (SLC5A3) gene and characterization of the promoter
Mallee J, et al.
Genomics, 46(3) (1997)
Human Na+-myo-inositol cotransporter gene: alternate splicing generates diverse transcripts
Porcellati F, et al.
American Journal of Physiology. Cell Physiology, 274(5) (1998)
Hypotonic activation of the myo-inositol transporter SLC5A3 in HEK293 cells probed by cell volumetry, confocal and super-resolution microscopy
Andronic J, et al.
PLoS ONE, 10(3), e0119990-e0119990 (2015)
Joseph Andronic et al.
PloS one, 10(3), e0119990-e0119990 (2015-03-11)
Swelling-activated pathways for myo-inositol, one of the most abundant organic osmolytes in mammalian cells, have not yet been identified. The present study explores the SLC5A3 protein as a possible transporter of myo-inositol in hyponically swollen HEK293 cells. To address this
The transport mechanism of the human sodium/myo-inositol transporter 2 (SMIT2/SGLT6), a member of the LeuT structural family
Sasseville L, et al.
American Journal of Physiology. Cell Physiology, 307(5) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service