Skip to Content
Merck
All Photos(1)

Key Documents

SAB1402047

Sigma-Aldrich

Anti-HTRA4 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ90724

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTRA4(203100)

General description

This gene encodes a member of the HtrA family of proteases. The encoded protein contains a putative signal peptide, an insulin growth factor binding domain, a Kazal protease inhibitor domain, a conserved trypsin domain and a PDZ domain. Based on studies on other related family members, this enzyme may function as a secreted oligomeric chaperone protease to degrade misfolded secretory proteins. Other human HtrA proteins have been implicated in arthritis, tumor suppression, unfolded stress response, apoptosis, and aging. (provided by RefSeq)

Immunogen

HTRA4 (AAH57765.1, 1 a.a. ~ 476 a.a) full-length human protein.

Sequence
MIRPQLRTAGLGRCLLPGLLLLLVPVLWAGAEKLHTQPSCPAVCQPTRCPALPTCALGTTPVFDLCRCCRVCPAAEREVCGGAQGQPCAPGLQCLQPLRPGFPSTCGCPTLGGAVCGSDRRTYPSMCALRAENRAARRLGKVPAVPVQWGNCGDTGTRSAGPLRRNYNFIAAVVEKVAPSVVHVQLWGRLLHGSRLVPVYSGSGFIVSEDGLIITNAHVVRNQQWIEVVLQNGARYEAVVKDIDLKLDLAVIKIESNAELPVLMLGRSSDLRAGEFVVALGSPFSLQNTATAGIVSTKQRGGKELGMKDSDMDYVQIDATINYGNSGGPLVNLDGDVIGVNSLRVTDGISFAIPSDRVRQFLAEYHEHQMKGKAFSNKKYLGLQMLSLTVPLSEELKMHYPDFPDVSSGVYVCKVFEGTAAQSSGLRDHDVIVNINGKPITTTTDVVKALDSDSLSMAVLRGKDNLLLTVIPETIN

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Renke et al.
Acta biochimica Polonica, 65(3), 471-478 (2018-08-28)
Mast cells play an important role in both, the innate and adaptive immunity, however, clonal proliferation of abnormal mast cells in various organs leads to mastocytosis. A skin variant of the disease, cutaneous mastocytosis (CM) is the most frequent form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service