Skip to Content
Merck
All Photos(1)

Key Documents

SAB1307254

Sigma-Aldrich

ANTI-BAT1(C-TERMINAL) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

56 kDa U2AF65-associated protein, BAT1, DDX39B, Spliceosome RNA helicase DDX39B, UAP56

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

48991 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX39B(7919)

Related Categories

General description

BAT1, also known as UAP56 or DExD-box helicase 39B (DDX39B), is encoded in the central region of the major histocompatibility complex (MHC) on the short arm of human chromosome 6p21.33. It is a member of DEAD box family of RNA-binding proteins and is characterized with the nine conserved motifs including ATPase and RNA helicase motifs.

Application

ANTI-BAT1(C-TERMINAL) antibody produced in rabbit has been used in immunoprecipitation and western blotting.

Biochem/physiol Actions

BAT1/ UAP56 facilitates the binding of U2 small nuclear ribonucleoprotein (SnRP) to pre-mRNA. It also helps in the export of mRNA from the nucleus to the cytoplasm. Spliceosome RNA helicase BAT1 is implicated in adapting nucleic acid metabolism to cellular stress. BAT1 is implicated in the down-regulation of inflammation. The gene polymorphism might be associated with the development of allergic diseases such as bronchial asthma, allergic rhinitis, atopic dermatitis, food allergy, drug allergy and pollen allergy. Mutation in the gene may also lead to the development of urticaria and Quincke′s edema.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M B Freĭdin et al.
Molekuliarnaia biologiia, 45(3), 464-472 (2011-07-28)
Genome-wide association studies are currently considered as one of the most powerful tools to establishing the genetic basis of complex diseases. A number of such studies were carried out for allergic diseases; however, in Russian population this analysis has not
Hang Shi et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(51), 17628-17633 (2004-12-09)
Pre-mRNA splicing requires the function of a number of RNA-dependent ATPases/helicases, yet no three-dimensional structure of any spliceosomal ATPases/helicases is known. The highly conserved DECD-box protein UAP56/Sub2 is an essential splicing factor that is also important for mRNA export. The
Influenza Virus mRNA Trafficking Through Host Nuclear Speckles.
Mor A, et al.
Nature microbiology, 2016 (2016)
EunMi Juliana Jung et al.
Proteomics, 7(21), 3906-3918 (2007-10-09)
Ambient particulate matter (PM) induces adverse health effects through the ability of pro-oxidative chemicals to induce the production of oxygen radicals and oxidant injury. Utilizing a proteomics strategy involving 2-D DIGE, immunoblotting, and real-time PCR, we demonstrate that organic diesel
R J Allcock et al.
Genes to cells : devoted to molecular & cellular mechanisms, 6(5), 487-494 (2001-05-31)
BAT1 belongs to the DEAD-box family of RNA-binding proteins and is encoded in the central MHC. To determine whether it affects immune responses and hence diseases influenced by MHC haplotypes, U937, THP1 and Jurkat cells were stably transfected with anti-sense

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service