Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305565

Sigma-Aldrich

MONOCLONAL ANTI-CD73(NT5E)(C-TERMINAL) antibody produced in mouse

clone 974CT7.5.3, crude ascites, buffered aqueous solution

Synonym(s):

5′-nucleotidase, NT5, NT5E, NTE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
974CT7.5.3, monoclonal
application:
WB
species reactivity:
human
technique(s):
western blot: 1:5000
citations:

biological source

mouse

Quality Level

antibody form

crude ascites

antibody product type

primary antibodies

clone

974CT7.5.3, monoclonal

form

buffered aqueous solution

mol wt

63368 Da

species reactivity

human

technique(s)

western blot: 1:5000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NT5E(4907)

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service