Skip to Content
Merck

Skip To

S9012

Sulforhodamine B sodium salt

Powder

Synonym(s):

Acid red 52; Acid Rhodamine B

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C27H29N2NaO7S2
CAS Number:
Molecular Weight:
580.65
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
EC Number:
222-529-8
MDL number:
Colour Index Number:
45100
Beilstein/REAXYS Number:
3886008
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Sulforhodamine B sodium salt, Technical grade

Quality Segment

form

powder

composition

Dye content, ~75%

εmax

≥85000 at 554-560 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccc(cc4S([O-])(=O)=O)S([O-])(=O)=O)=[N+](\CC)CC)c1

InChI

1S/C27H30N2O7S2.Na/c1-5-28(6-2)18-9-12-21-24(15-18)36-25-16-19(29(7-3)8-4)10-13-22(25)27(21)23-14-11-20(37(30,31)32)17-26(23)38(33,34)35;/h9-17H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1

InChI key

SXQCTESRRZBPHJ-UHFFFAOYSA-M

General description

Sulforhodamine B (also known as SRB, kiton rhodamine B, lissamine rhodamine B, or sulphorhodamine B) is a bright-pink anionic, aminoxanthene dye carrying two sulfonate substituents.

Application

Sulforhodamine B (SRB) is known for use in SRB assay for the determination of in vitro cytotoxicity through staining and the observation of live cells, proliferation and drug screening.

It has been used in the SRB assay to evaluate the effect of mouthwashes on protein content in primary cultures of human fibroblasts.

It is also suitable as a microscopic tracer for the assessment of blood-brain barrier integrity by post mortem inspection of frozen sections. SRB is useful as a caries detector dye with subsequent inspection of histological material.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S1402230162S7635
description

Technical grade

description

powder, BioReagent, suitable for cell culture

description

Dye content 75 %

description

Dye content ~95 %

form

powder

form

powder

form

powder

form

powder

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

-

application(s)

diagnostic assay manufacturing
hematology
histology

application(s)

diagnostic assay manufacturing
hematology
histology

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

composition

Dye content, ~75%

composition

Dye content, ~75%

composition

Dye content, 75%

composition

Dye content, ~95%

storage temp.

room temp

storage temp.

-

storage temp.

room temp

storage temp.

room temp

εmax

≥85000 at 554-560 nm

εmax

-

εmax

-

εmax

-


Still not finding the right product?

Explore all of our products under Sulforhodamine B sodium salt


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters