Skip to Content
Merck
All Photos(1)

Key Documents

S8451

Sigma-Aldrich

Saquinavir mesylate

≥98% (HPLC), powder

Synonym(s):

(2S)-N1[(1S,2R)-3-[(3S,4aS,8aS)-3-[[(1,1-Dimethylethyl)amino]carbonyl]octahydro-2(1H)-isoquinolinyl]-2-hydroxy-1-(phenylmethyl)propyl]-2-[(2-quinolinylcarbonyl)amino]butanediamide methanesulfonate, Ro-31-8959

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H50N6O5·CH4O3S
CAS Number:
Molecular Weight:
766.95
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥5 mg/mL

originator

Roche

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.CC(C)(C)NC(=O)[C@@H]1C[C@@H]2CCCC[C@@H]2CN1C[C@@H](O)[C@H](Cc3ccccc3)NC(=O)[C@H](CC(N)=O)NC(=O)c4ccc5ccccc5n4

InChI

1S/C38H50N6O5.CH4O3S/c1-38(2,3)43-37(49)32-20-26-14-7-8-15-27(26)22-44(32)23-33(45)30(19-24-11-5-4-6-12-24)41-36(48)31(21-34(39)46)42-35(47)29-18-17-25-13-9-10-16-28(25)40-29;1-5(2,3)4/h4-6,9-13,16-18,26-27,30-33,45H,7-8,14-15,19-23H2,1-3H3,(H2,39,46)(H,41,48)(H,42,47)(H,43,49);1H3,(H,2,3,4)/t26-,27+,30-,31-,32-,33+;/m0./s1

InChI key

IRHXGOXEBNJUSN-YOXDLBRISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Saquinavir is a peptidomimetic hydroxyethylamine inhibitor, which is used to treat human immunodeficiency virus (HIV) infection.

Application

Saquinavir mesylate may be used in cell signaling and immunology studies.

Biochem/physiol Actions

Saquinavir is an HIV Protease Inhibitor used in antiretroviral therapy.
Saquinavir is an HIV Protease Inhibitor used in antiretroviral therapy. It inhibits both HIV-1 and HIV-2 proteases. Studies have also looked at saquinavir as a possible anti-cancer agent.

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound was developed by Roche. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Davis et al.
Antimicrobial agents and chemotherapy, 56(7), 3620-3628 (2012-04-18)
Inhibitors of HIV protease have proven to be important drugs in combination anti-HIV therapy. These inhibitors were designed to target mature protease and prevent viral particle maturation by blocking Gag and Gag-Pol processing by mature protease. Currently there are few
Martha Stefanidou et al.
Antimicrobial agents and chemotherapy, 56(8), 4381-4390 (2012-06-06)
The maturation of newly formed human immunodeficiency virus type 1 (HIV-1) virions is a critical step for the establishment of productive infection. We investigated the potential of saquinavir (SQV), a protease inhibitor (PI) used in highly active antiretroviral therapy (HAART)
Can inhibition of proteasomes or NF-kappaB help control idiopathic nephrotic syndrome?
Scott E Wenderfer
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 27(5), 1698-1701 (2012-05-02)
Desk Reference of Clinical Pharmacology (2007)
Zhiying Wang et al.
Journal of pharmaceutical sciences, 101(9), 3199-3213 (2012-05-23)
In an approach to overcome biological barriers mediated by P-glycoprotein (P-gp) and cytochrome P450 3A (CYP3A), a series of stereoisomeric valine-valine prodrugs of saquinavir (SQV) were synthesized and investigated with respect to affinity for efflux pump P-gp, and resistance to

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service