Skip to Content
Merck
All Photos(1)

Key Documents

S7776

Sigma-Aldrich

Starch Azure

Insoluble corn starch covalently linked with Remazol Brilliant Blue R

Synonym(s):

Starch–Remazol brilliant blue R, RBB-Starch, Remazol brilliant blue R dyed starch, Starch Azure

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352204
NACRES:
NA.28

Quality Level

form

powder

Looking for similar products? Visit Product Comparison Guide

General description

Starch is an insoluble starch substrate specific for α-amylase and not for other exo enzymes. α-amylase catalysis of starch azure solubilization is measured at 595 nm.

Application

Starch Azure has been used as a substrate
  • in the screening of α-amylase activity in decontaminated malted flour at varying pH and temperature
  • for monitoring the inhibitory activity of Physalis peruviana extracts
  • for thermostable α-amylase extracted from deep sea and acidic soil

Substrates

Substrate for colorimetric determination of α-amylase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A novel, high performance enzyme for starch liquefaction discovery and optimization of a low pH, thermostable alpha-amylase
Richardson TH, et al.
The Journal of Biological Chemistry, 277(29), 26501-26507 (2002)
Inhibitory effects of an extract of fruits of Physalis peruviana on some intestinal carbohydrases
Rey D, et al.
Revista Colombiana de Ciencias Quimico-Farmaceuticas, 44(1), 72-89 (2015)
The use of starch azure for measurement of alpha-amylase activity
Lehoczki G, et al.
Carbohydrate Polymers, 183, 263-266 (2018)
Modeling cereal starch hydrolysis during simultaneous saccharification and lactic acid fermentation; case of a sorghum-based fermented beverage, gowe
Mestres C, et al.
Food Research International, 100, 102-111 (2017)
Gábor Lehoczki et al.
Carbohydrate polymers, 183, 263-266 (2018-01-22)
Despite being widely used, there is no standard protocol for α-amylase activity measurement with starch azure substrate. Boiling pre-treatment of starch azure suspension increased the reaction rate of hydrolysis catalysed by human salivary α-amylase (HSA) or porcine pancreatic α-amylase (PPA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service