Skip to Content
Merck
All Photos(3)

Key Documents

S3126

Sigma-Aldrich

Sperm nuclei from salmon

powder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

product name

Sperm nuclei from salmon, Type II-S

biological source

fish (salmon)

type

Type II-S

form

powder

storage temp.

−20°C

Biochem/physiol Actions

An excellent source of protamine and high molecular weight DNA.

Quality

Crude

Preparation Note

Carefully processed to minimize degradation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc-André Déry et al.
Scientific reports, 7, 42285-42285 (2017-02-17)
The cellular prion protein (PrP) is essential for transmissible prion diseases, but its exact physiological function remains unclear. Better understanding the regulation of the human prion protein gene (PRNP) expression can provide insight into this elusive function. Spliced XBP1 (sXBP1)
T Samuel
Clinical and experimental immunology, 32(2), 290-298 (1978-05-01)
The indirect immunofluorescence test on swollen nuclei of rat thymocytes, chicken red blood cells and human and salmon spermatozoa was found to be an easy and satisfactory method for the discrimination between antibodies to sperm-specific nuclear antigens and somatic nuclear
Yuguo Zang et al.
PloS one, 6(9), e25282-e25282 (2011-10-04)
Crystallization conditions of an intact monoclonal IgG4 (immunoglobulin G, subclass 4) antibody were established in vapor diffusion mode by sparse matrix screening and subsequent optimization. The procedure was transferred to microbatch conditions and a phase diagram was built showing surprisingly
M D Myers et al.
The New phytologist, 132(3), 445-451 (1996-03-01)
Mycorrhizal plants of Vaccinium mocrocarpon Aiton used nuclei from salmon sperm as a sole source of phosphorus (P) and achieved similar yields, P content and P concentration to plants crown with orthophosphate. Mycorrhizal infection significantly increased the effectiveness of utilization
A G Balliet et al.
DNA and cell biology, 20(4), 239-247 (2001-06-14)
The MyD118 (Gadd45beta) protein is a member of a family of structurally related proteins, including Gadd45 (Gadd45alpha) and CR6 (Gadd45gamma), that have critical roles in regulating growth arrest and apoptosis. The MyD118 and other members of its family display distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service