Skip to Content
Merck
All Photos(4)

Key Documents

S3062

Sigma-Aldrich

Anti-α-Synuclein antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-SNCA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 19 kDa

species reactivity

rat, human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200 using formic acid treated sections of Alzheimer′s Disease brain
microarray: suitable
western blot: 1:1,000 using extract of rat brain homogenate

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)
rat ... Snca(29219)

General description

The synucleins are a family of soluble presynaptic proteins that are abundant in neurons and include α-synuclein, β-synuclein, and γ-synuclein. Human α-synuclein (also known as the non-amyloid component of plaques precursor protein or NACP) is a 140-amino acid polypeptide that is encoded by a gene on chromosome 4. It was originally isolated from plaques of Alzheimer′s disease (AD) brains as a 19 kDa protein precursor of the highly hydrophobic 35-amino acid peptide, nonamyloid component (NAC) of plaques. The NAC can self-aggregate into fibrils and induce aggregation of the β-amyloid peptide. α-Synuclein is highly abundant in presynaptic terminals, and is a major component of Lewy bodies (LBs). Pathogenic point mutations in the α-synuclein gene are linked to familial Parkinson′s disease (PD) in rare kindreds. However, most neurodegenerative disorders with LBs are associated with abnormal accumulation of wild-type, α-synuclein. α-Synuclein is expressed primarily in brain, but is also expressed in low levels in all tissues examined except liver.

Specificity

Rabbit polyclonal anti-α-Synuclein antibody recognizes human and rat α-synuclein. Staining of α-synuclein in immunoblotting is specifically inhibited by the immunizing peptide

Immunogen

synthetic peptide corresponding to a sequence near the C-terminus of human α-synuclein (amino acids 111-132 with C-terminally added lysine) conjugated to KLH. This sequence is highly conserved (90% homology) in mouse and rat α-synuclein. This sequence has no homology with β- and γ-synuclein.

Application

Rabbit polyclonal anti-α-Synuclein antibody is used to tag α-Synuclein for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques, such as immunoblotting. It is also used as a probe to determine the presence and roles of α-Synuclein in plaques of Alzheimer′s disease (AD) brains.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Identification of a caspase-derived N-terminal tau fragment in cellular and animal Alzheimer's disease models
Corsetti V, et al.
Molecular and Cellular Neurosciences, 38(3), 381-392 (2008)
Extracellular truncated tau causes early presynaptic dysfunction associated with Alzheimer?s disease and other tauopathies
Florenzano F, et al.
Oncotarget, 8(39), 64745-64778 (2017)
Clinical, Histological, and Immunohistochemical Findings in Inclusion Body Myositis
Camargo L, et al.
BioMed Research International, 2018 (2018)
Lysosomal dysfunction in the brain of a mouse model with intraneuronal accumulation of carboxyl terminal fragments of the amyloid precursor protein
Kaur G, et al.
Molecular Psychiatry, 22(7), 981-981 (2017)
Geert Callewaert et al.
Frontiers in genetics, 11, 266-266 (2020-05-28)
The yeast Saccharomyces cerevisiae is a powerful model to study the molecular mechanisms underlying α-synuclein (α-syn) cytotoxicity. This is due to the high degree of conservation of cellular processes with higher eukaryotes and the fact that yeast does not endogenously

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service