Skip to Content
Merck
All Photos(1)

Key Documents

S2147

Sigma-Aldrich

Monoclonal Anti-Superoxide Dismutase (SOD) antibody produced in mouse

clone SD-G6, ascites fluid

Synonym(s):

Anti-ALS, Anti-ALS1, Anti-HEL-S-44, Anti-IPOA, Anti-SOD, Anti-STAHP, Anti-hSod1, Anti-homodimer

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

SD-G6, monoclonal

contains

15 mM sodium azide

species reactivity

canine, human, rat

technique(s)

indirect ELISA: 1:300

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOD1(6647)
rat ... Sod1(24786)

General description

Monoclonal Anti-Superoxide Dismutase (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. Superoxide Dismutase (SOD) is a family of metalloenzymes widely distributed in both plants and animals. In mammalian tissues, three types of superoxide dismutase [Cu-Zn-SOD, Mn-SOD, extracellular (EC)-SOD] occur. Human manganese superoxide dismutase (MnSOD), isolated from liver is composed of 22 kDa subunits each containing one Mn atom, while SOD from bovine erythrocyte has a molecular weight of 32.5 kDa. Superoxide Dismutase occurs in high concentrations in brain, liver, heart, erythrocytes and kidney.
Superoxide Dismutase (SOD) or CuZn-SOD (SOD1), a cytoplasmic and mitochondrial intermembrane space protein is located on human chromosome 21q22. It belongs to superoxide dismutase multigene family.

Specificity

The antibody recognizes natural and recombinant human-Cu-Zn-SOD, human placental SOD, and human erythrocyte SOD using direct capture or competitive ELISA. Cross-reactivity has been observed with human liver and salivary gland, rat salivary gland, pheochromocytoma cell line (PC12), and dog salivary gland. No reactivity was observed with SOD from bovine erythrocytes, kidney, and liver; dog erythrocytes; Bacillus stearothermophilus; E. coli, or horseradish.

Immunogen

recombinant human copper-zinc superoxide dismutase (Cu-Zn-SOD).

Application

Anti-Superoxide Dismutase (SOD) antibody has been used in immunohistochemistry and Cu-Zn SOD detection via ELISA.

Biochem/physiol Actions

Superoxide Dismutase (SOD) appear to protect cells against reactive free radicals by scavenging the superoxide radicals produced by ionization radiation or through other mechanisms. SOD have been proposed as clinically useful for a wide variety of applications including prevention of oncogenesis, tumor promotion, tumor invasiveness, radiation damage, reduction of the cytotoxic and cardiotoxic effects of anticancer drugs, as a measure against the aging process and as anti-inflammatory agents.
Superoxide Dismutase (SOD) or CuZn-SOD (SOD1) mutations results in amyotrophic lateral sclerosis It acts as a mediator of the HMF (hypomagnetic field) effect.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Functional Role of MnSOD as a Biomarker of Human Diseases and Therapeutic Potential of a New Isoform of a Human Recombinant MnSOD
Borrelli A, et al.
BioMed Research International, 2014 (2014)
Superoxide Dismutases: Role in Redox Signaling, Vascular Function, and Diseases
Fukai T, et al.
Antioxidants & Redox Signaling, 15(6), 1583-1583 (2011)
Marcel Maier et al.
Science translational medicine, 10(470) (2018-12-07)
Mutations in the gene encoding superoxide dismutase 1 (SOD1) lead to misfolding and aggregation of SOD1 and cause familial amyotrophic lateral sclerosis (FALS). However, the implications of wild-type SOD1 misfolding in sporadic forms of ALS (SALS) remain unclear. By screening
Targeted expression of human CuZn superoxide dismutase gene in mouse central nervous system
Li Y, et al.
Journal of Neuroscience Methods, 89(1), 49-55 (1999)
Superoxide dismutase multigene family: a comparison of the CuZn-SOD (SOD1), Mn-SOD (SOD2), and EC-SOD (SOD3) gene structures, evolution, and expression
Zelko IN, et al.
Free Radical Biology & Medicine, 33(3), 337-349 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service