Skip to Content
Merck
All Photos(1)

Documents

S179

Sigma-Aldrich

R(+)-SKF-81297 hydrobromide

≥98% (HPLC), solid

Synonym(s):

R-(+)-6-Chloro-7,8-dihydroxy-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16ClNO2 · HBr
CAS Number:
Molecular Weight:
370.67
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

optical activity

[α]22/D +15.14°, c = 0.52 in DMF(lit.)

storage condition

desiccated
protect from light

color

white to off-white

solubility

H2O: 6 mg/mL
DMSO: soluble

SMILES string

Br[H].Oc1cc2[C@H](CNCCc2c(Cl)c1O)c3ccccc3

InChI

1S/C16H16ClNO2.BrH/c17-15-11-6-7-18-9-13(10-4-2-1-3-5-10)12(11)8-14(19)16(15)20;/h1-5,8,13,18-20H,6-7,9H2;1H/t13-;/m1./s1

InChI key

RMIJGBMRNYUZRG-BTQNPOSSSA-N

Application

R(+)-SKF-81297 hydrobromide has used as D1-type dopamine receptor agonist:
  • to test it effect on the spike firing in rat retinal ganglion cells
  • in human embryonic kidney (HEK) 293T cells
  • to test its inhibitory effect in microglial cells

Biochem/physiol Actions

SKF-81297 administration in spontaneously hypertensive rats (SHR) induces an increase in expression of proto-oncogene c-fos mRNA leading to biphasic effect on locomotion. It favors the turnover of phosphoinositide and adenylyl cyclase. SKF-81297 is involved in the stimulation of neurons, in particular the postsynaptic striatal neurons.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rochellys Diaz Heijtz et al.
Behavioral and brain functions : BBF, 2, 18-18 (2006-05-30)
Molecular genetic studies suggest the dopamine D1 receptor (D1R) may be implicated in attention-deficit/hyperactivity disorder (ADHD). As little is known about the potential motor role of D1R in ADHD, animal models may provide important insights into this issue. We investigated
Maximilian Stalter et al.
Cell reports, 30(1), 164-172 (2020-01-09)
The neurotransmitter dopamine, which acts via the D1-like receptor (D1R) and D2-like receptor (D2R) family, may play an important role in gating sensory information to the prefrontal cortex (PFC). We tested this hypothesis in awake macaques and recorded visual motion-direction
R J Vermeulen et al.
European journal of pharmacology, 235(1), 143-147 (1993-04-22)
The alleged selective, high efficacy dopamine D1 receptor agonist, SKF 81297 (0.05-0.3 mg/kg i.m.), induced rotational behaviour away from the lesion and stimulated use of the dominant right hand in unilaterally (left side) 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-lesioned rhesus monkeys (Macaca mulatta). The
C. Reavill et al.
Behavioural pharmacology, 4(2), 135-146 (1993-04-01)
A range of selective dopamine D1 and D2 receptor agonists and antagonists was used to characterize to the discriminative stimuli produced by d-amphetamine (0.5mg/kg) and the D1 agonists SKF 81297 (0.1mg/kg). In rats trained to discriminate d-amphetamine (0.5mg/kg) from saline
J Bergman et al.
The Journal of pharmacology and experimental therapeutics, 273(1), 40-48 (1995-04-01)
The effects of dopamine agonists differing in affinity and selectivity at D1 and D2 types of dopamine receptors were compared in squirrel monkeys responding under two different schedules of reinforcement: a fixed-interval (FI) schedule of stimulus-shock termination and a fixed-ratio

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service