Skip to Content
Merck
All Photos(1)

Key Documents

S1697

Sigma-Aldrich

SPD304

≥98% (HPLC), solid

Synonym(s):

6,7-Dimethyl-3-[[methyl[2-[methyl[[1-[3-(trifluoromethyl)phenyl]-1H-indol-3-yl]methyl]amino]ethyl]amino]methyl]-(4H-1-Benzopyran-4-one dihydrochloride, SPD00000304

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H32F3N3O2 · 2HCl
CAS Number:
Molecular Weight:
620.53
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

Cl.Cl.CN(CCN(C)Cc1cn(-c2cccc(c2)C(F)(F)F)c3ccccc13)CC4=COc5cc(C)c(C)cc5C4=O

InChI

1S/C32H32F3N3O2.2ClH/c1-21-14-28-30(15-22(21)2)40-20-24(31(28)39)18-37(4)13-12-36(3)17-23-19-38(29-11-6-5-10-27(23)29)26-9-7-8-25(16-26)32(33,34)35;;/h5-11,14-16,19-20H,12-13,17-18H2,1-4H3;2*1H

InChI key

GOZMBJCYMQQACI-UHFFFAOYSA-N

Gene Information

human ... TNF(7124)
mouse ... TNF(21926)
rat ... TNF(24835)

Application

SPD304 has been used in the inhibition of tumor necrosis factor receptor 1 (TNFR1) in dorsal root ganglion neurons and tumor necrosis factor alpha in colonic epithelial cell lines.
SPD304 may be used in TNF-α-mediated cell signaling studies.

Biochem/physiol Actions

SPD304 is a small molecule inhibitor of TNF-a activity with a novel mechanism of action. It interferes with protein-protein interactions on the contact surfaces of the trimeric TNF-a subunits, displacing one of the trimer units, resulting in dissociation of the trimer and loss of activity at the receptor TNFR1. SPD304 inhibits TNFR1 receptor binding to TNF-a with an IC50 of 22μM in vitro and inhibits TNF-a-mediated stimulation of IkB degradation in HeLa cells with an IC50 of 4.6 μM.
Trimerization of TNF-α is essential for its biological activity. SPD304 inhibits the trimerization by interacting with Glycine 122 residue. It also inhibits the activity of Receptor activator of nuclear factor-κB ligand (RANKL) by similar interaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cannabinoid WIN-55,212-2 mesylate inhibits tumor necrosis factor-alpha-induced expression of nitric oxide synthase in dorsal root ganglion neurons
Tan R and Cao L
International Journal of Molecular Medicine, 42(2), 919-925 (2018)
Molly M He et al.
Science (New York, N.Y.), 310(5750), 1022-1025 (2005-11-15)
We have identified a small-molecule inhibitor of tumor necrosis factor alpha (TNF-alpha) that promotes subunit disassembly of this trimeric cytokine family member. The compound inhibits TNF-alpha activity in biochemical and cell-based assays with median inhibitory concentrations of 22 and 4.6
Dysregulated Up-Frameshift Protein 1 Promotes Ulcerative Colitis Pathogenesis Through the TNFR1-NF-kappaB/MAPKs Pathway
Zhu H, et al.
Digestive Diseases and Sciences, 63(10), 2593-2603 (2018)
Yang Yu et al.
Hypertension (Dallas, Tex. : 1979), 74(1), 63-72 (2019-06-04)
TNF-α (tumor necrosis factor-α) is initially synthesized as a transmembrane protein that is cleaved by TACE (TNF-α-converting enzyme) to release soluble TNF-α. The elevated level of TNF-α in the brain and circulation in heart failure (HF) suggests an increase in
Yan Zhang et al.
Molecular cancer research : MCR, 12(8), 1181-1191 (2014-05-14)
The relationship between tumor-associated macrophages (TAM) and epithelial-to-mesenchymal transition (EMT) during the initiation and progression of metastasis is still unclear. Here, a role for the vitamin D receptor (VDR) in metastasis was identified, as well as a role in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service