Skip to Content
Merck
All Photos(1)

Key Documents

S1076

Sigma-Aldrich

SRC, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

ASV, c-SRC, p60-Src

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in E. coli

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

83-113 nmol/min·mg

mol wt

~83 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SRC(6714)

General description

SRC proto-oncogene, non-receptor tyrosine kinase (SRC) belongs to the family of non-receptor tyrosine kinases. SRC was originally identified as a transforming protein of the Rous sarcoma virus (RSV) that had enzymatic ability to phosphorylate tyrosine in protein substrates. It contains an amino-terminal domain, conserved Src-homology domains (SH2 and SH3), a tyrosine kinase domain and the carboxyl-terminus. The gene encoding this protein is localized on human chromosome 20q11.23.

Application

SRC, active, GST tagged human has been used in the in vitro kinase assay.

Biochem/physiol Actions

SRC proto-oncogene, non-receptor tyrosine kinase (SRC) phosphorylates membrane proteins and nucleoproteins and activates various intracellular signaling cascades. It is overexpressed and activated in a large number of human malignancies and has been linked to the development of cancer and progression to distant metastases. In addition to increasing cell proliferation, a key role of SRC in cancer seems to be the ability to promote invasion and motility, functions that might contribute to tumor progression.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fangfang Chen et al.
American journal of cancer research, 8(8), 1466-1482 (2018-09-14)
R428 (BGB324) is an anti-cancer drug candidate under clinical investigation. It inhibits the receptor tyrosine kinase Axl and induces apoptosis of many types of cancer cells, but the relationship between the two has not been well established. We investigated the
Min Yu et al.
Microbiology and immunology, 65(1), 40-47 (2020-10-08)
The ischemia-reperfusion-induced damage in human brain microvascular endothelial cells (BMECs) is associated with disruption of the blood-brain barrier. CXC chemokine ligand 5 (CXCL5) is reported to be up-regulated in ischemic stroke. However, the detailed function of CXCL5 in this pathological
Yun Zhou et al.
International journal of ophthalmology, 13(12), 1887-1894 (2020-12-22)
To investigate the effects and mechanism of β-elemene on the expressions of hypoxia-inducible factor-1α (HIF-lα), vascular endothelial growth factor (VEGF) and inducible nitric oxide synthase (iNOS) in human retinal pigment epithelial (RPE) cells under high glucose conditions. ARPE-19 cell line
M S Collett et al.
Proceedings of the National Academy of Sciences of the United States of America, 75(4), 2021-2024 (1978-04-01)
Incorporation of phosphorus from [gamma-32P]ATP into protein was catalyzed by specific immunoprecipitates from avian sarcoma virus (ASV)-transformed avian and mammalian cells. This incorporation was observed only when antiserum from tumor-bearing rabbits able to specifically precipitate the ASV sarcoma gene product
Genomic imbalances in Schistosoma-associated and non-Schistosoma-associated bladder carcinoma. An array comparative genomic hybridization analysis.
Armengol G
Cancer Genetics and Cytogenetics, 177(1), 16-19 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service