Skip to Content
Merck
All Photos(1)

Key Documents

S0693

Sigma-Aldrich

SB 204741

>98% (HPLC)

Synonym(s):

N-(1-Methyl-1H-5-indolyl)-N′-(3-methyl-5-isothiazolyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4OS
CAS Number:
Molecular Weight:
286.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

>98% (HPLC)

form

solid

color

off-white to tan

solubility

DMSO: soluble >20 mg/mL

originator

GlaxoSmithKline

storage temp.

2-8°C

SMILES string

Cc1cc(NC(=O)Nc2ccc3n(C)ccc3c2)sn1

InChI

1S/C14H14N4OS/c1-9-7-13(20-17-9)16-14(19)15-11-3-4-12-10(8-11)5-6-18(12)2/h3-8H,1-2H3,(H2,15,16,19)

InChI key

USFUFHFQWXDVMH-UHFFFAOYSA-N

Application

SB 204741 has been used as a 5-hydroxytryptamine receptor 2B (Htr2B) antagonist:
  • to inhibit Htr2b signaling to test its effect on hepatic stellate cell (HSC) activation in three-month-old adult zebrafish
  • to test its effect on the inhibition of fluoxetine effects in depressive-like behavioral studies in mice
  • to test its effect in reducing scratching behavior evoked by sertraline in mice

Biochem/physiol Actions

SB 204741 reduces the synaptic facilitation induced by the low dose of buprenorphine in spinal C-fiber synapses.
5-HT2B serotonin receptor antagonist.

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peripheral serotonin receptor 2B and transient receptor potential channel 4 mediate pruritus to serotonergic antidepressants in mice.
Sang Hoon Lee et al.
The Journal of allergy and clinical immunology, 142(4), 1349-1352 (2018-06-20)
Ercan Ozdemir et al.
Bioelectromagnetics, 40(5), 319-330 (2019-06-04)
Much evidence demonstrates the antinociceptive effect of magnetic fields (MFs). However, the analgesic action mechanism of the electromagnetic field (EMF) is not exactly understood. The aim of the present study was to investigate the effects of 5-HT1 and 5-HT2 receptor
Qiqi Yang et al.
Cellular and molecular gastroenterology and hepatology, 3(3), 484-499 (2017-05-04)
Hepatocellular carcinoma (HCC) occurs more frequently and aggressively in men than in women. Although sex hormones are believed to play a critical role in this disparity, the possible contribution of other factors largely is unknown. We aimed to investigate the
Xiaowei Li et al.
Frontiers in psychiatry, 9, 734-734 (2019-01-23)
The long-lasting loss of sleep is a generally acknowledged risk factor for the occurrence of major depressive disorder (MDD), whereas sleep abnormalities being a key clinic symptom of the MDD. In our previous work, we demonstrated that the sleep deprivation
Annika Gocht et al.
Transplant immunology, 49, 43-53 (2018-04-13)
Cardiac allograft vasculopathy (CAV) is the main obstacle for long-term survival after heart transplantation. Alloimmune mediated chronic vascular rejection results in several mechanisms like platelet activation, immigration of inflammatory cells through the endothelial layer and proliferation and migration of smooth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service