Skip to Content
Merck
All Photos(1)

Key Documents

S009

Sigma-Aldrich

LY-165,163

solid

Synonym(s):

4-[2-[4-[3-(Trifluoromethyl)phenyl]-1-piperazinyl]ethyl]benzeneamine, p-Aminophenethyl-m-trifluoromethylphenyl piperazine, PAPP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H22F3N3
CAS Number:
Molecular Weight:
349.39
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

color

off-white

solubility

0.1 M HCl: slightly soluble
H2O: insoluble
ethanol: freely soluble

SMILES string

Nc1ccc(CCN2CCN(CC2)c3cccc(c3)C(F)(F)F)cc1

InChI

1S/C19H22F3N3/c20-19(21,22)16-2-1-3-18(14-16)25-12-10-24(11-13-25)9-8-15-4-6-17(23)7-5-15/h1-7,14H,8-13,23H2

InChI key

GAAKALASJNGQKD-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Selective 5-HT1A serotonin receptor agonist and 5-HT1D serotonin receptor antagonist.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P H Hutson et al.
European journal of pharmacology, 143(2), 221-228 (1987-11-10)
The putative 5-HT1A agonist 1-[2-(4-aminophenyl)ethyl]-4-(3-trifluoromethylphenyl)piperazine (LY165163, PAPP) (1, 2, 4, 10 mg/kg s.c.) caused a significant and dose-dependent hypothermia in rats, 30 and 60 min after injection. The decreases of temperature were less marked than that caused by 8-OH-DPAT 1
T P Donohoe et al.
Psychopharmacology, 93(1), 82-86 (1987-01-01)
The putative serotonin (5-HT)1A agonist 1-[2-(4-aminophenyl)ethyl]-4-(3-trifluormethylphenyl) piperazine (LY165163, PAPP) induces hyperphagia and hypothermia in rats, but unlike other 5-HT agonists, does not induce 5-HT stereotypy even at high doses (10 mg/kg sc). LY165163 (1 mg/kg) increased striatal DOPA accumulation in
M J Millan et al.
The Journal of pharmacology and experimental therapeutics, 269(1), 121-131 (1994-04-01)
In view of the involvement of central alpha 1-adrenoceptors in the expression of 5-HT1A receptor-mediated spontaneous tail-flicks (STFs) in the rat, this study examined whether the putative alpha 1-adrenoceptor antagonist (alpha 1-antagonist) properties of certain 5-HT1A receptor agonists, (+)-flesinoxan and
H C Jackson et al.
Neuropharmacology, 28(6), 635-642 (1989-06-01)
The effects of a variety of 5-hydroxytryptamine (5-HT) receptor agonists and antagonists on behaviour in 5- and 20-day old rat pups have been investigated. Increased locomotion and head-weaving responses were induced in both age groups by 8-OH-DPAT (8-hydroxy-2-(di-n-propylamino)tetralin; 5-HT1A agonist);
R B McCall et al.
The Journal of pharmacology and experimental therapeutics, 242(3), 1152-1159 (1987-09-01)
Previous studies indicate that serotonin (5-HT) neurons provide a tonic excitatory input to central sympathetic neurons. The purpose of the present study was to utilize a number of 5-HT agonists in order to provide insights into the general function of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service