RTN10
GenElute™ Mammalian Total RNA Miniprep Kit
sufficient for 10 purifications
Synonym(s):
GenElute™ Mammalian RNA Kit, Gen Elute
About This Item
Recommended Products
usage
sufficient for 10 purifications
greener alternative product characteristics
Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
greener alternative category
, Aligned
storage temp.
15-25°C
Looking for similar products? Visit Product Comparison Guide
General description
denaturation of macromolecules and inactivation of RNases. Addition of ethanol causes RNA to bind when the lysate is spun through a silica membrane in a microcentrifuge tube. After washing to remove contaminants, RNA is eluted in 50-100 μL of Elution Solution. Up to 150 μg of total RNA can be isolated in less than 30 minutes.
If all traces of DNA contamination must be eliminated, further treatment with DNase I is recommended. DNase I digestion can be performed while the RNA is bound to the GenElute Binding Column using the On-Column DNase I Digestion Set (DNASE10 and DNASE70). Alternatively, for more stringent removal of contaminating DNA, the final RNA preparation can be treated with Amplification Grade DNase I (AMPD1).
Application
Features and Benefits
- Purifies total RNA from up to 107 cells or 40 mg of tissue per prep
- Yields up to 150 μg of pure, concentrated total RNA per prep
- Recover RNA from as few as 100 cells
- Simple and efficient–12 to 18 preps in 30 minutes
- Faster than gravity flow anion exchange methods
- No cumbersome steps associated with resins and magnetic slurries
- 40% more purifications per kit than the leading supplier
- Yields up to 150 μg of pure, concentrated total RNA per prep
- Purifies total RNA from up to 107 cells or 40 mg of tissue per prep
- Recover RNA from as few as 100 cells
Other Notes
Principle
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3
Target Organs
Central nervous system, Liver,Heart
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
Flash Point(F)
77.0 °F - closed cup
Flash Point(C)
25 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
profile of the IGF1 gene in Malabari and Attappady Black goats of India
Protocols
Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.
Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.
Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.
Learn Northern and Southern blotting basics, with protocols and applications for macromolecule transfer to membrane supports.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service