RAB1075
Human Periostin ELISA Kit
for cell culture supernatants, plasma, and serum samples
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
input
sample type serum sample(s)
sample type cell culture supernatant(s)
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 80 pg/ml
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... POSTN(10631)
General description
Periostin is an osteoblast-specific factor 2 and fasciclin-containing protein family member.
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.
Please refer to the attached Protocolfor details.
Human Periostin ELISA Kit has been used in the measurement of periostin in serum samples of atopic dermatitis and aneurysmal subarachnoid hemorrhage (aSAH) patients.
Biochem/physiol Actions
Periostin is crucial for the development of bone, tooth, and heart valves as well as in the synthesis of the extracellular matrix.
Other Notes
A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Myongsoon Sung et al.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology, 28(6), 543-550 (2017-06-21)
Serum periostin might be a biomarker in the pathogenesis of T helper 2-type allergic diseases. The aim of this study was to investigate the relationship between serum periostin levels and the severity and chronicity of atopic dermatitis (AD) in children.
Evaluation of serum periostin level in atopic dermatitis patients
Ghanem AH, et al.
Al-Azhar Assiut Medical Journal, 15(4), 223-223 (2017)
J A Castro-Rodriguez et al.
Allergologia et immunopathologia, 46(3), 235-240 (2017-12-28)
In contrast to adult asthmatic patients, studies on the role of serum periostin levels in schoolchildren with asthma are still conflictive, and very few studies have been performed in pre-schoolers. The aim of this study was to compare serum periostin
José M Rodrigo-Muñoz et al.
Allergy, 74(3), 507-517 (2018-07-25)
Asthma is a syndrome characterized by airway inflammation and obstruction. Due to its heterogeneity, the difficulties in asthma diagnosis and treatment make the discovery of new biomarkers a focus of research. So, we determined the differential miRNA expression of eosinophils
Diagnostic accuracy of T2 biomarkers for the prediction of airway eosinophilia in treated smoking asthmatic patients with loss of asthma control.
Evgenia Papathanasiou et al.
The journal of allergy and clinical immunology. In practice, 8(1), 385-387 (2019-07-02)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service