Skip to Content
Merck
All Photos(1)

Documents

RAB0581

Sigma-Aldrich

Cotton Rat Ifng / Interferon Gamma ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

cotton rat

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

The antibody pair provided in this kit recognizes Cotton rat IFN-gamma.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erina Petrera et al.
BioMed research international, 2014, 947560-947560 (2014-08-26)
Polyfunctionalized stigmasterol derivatives, (22S,23S)-22,23-dihydroxystigmast-4-en-3-one (compound 1) and (22S,23S)-3β-bromo-5α,22,23-trihydroxystigmastan-6-one (compound 2), inhibit herpes simplex virus type 1 (HSV-1) replication and spreading in human epithelial cells derived from ocular tissues. Both compounds reduce the incidence and severity of lesions in a murine
Linda A Lieberman et al.
PloS one, 9(10), e111382-e111382 (2014-11-02)
Systemic lupus erythematosus (SLE) is characterized by multiple cellular abnormalities culminating in the production of autoantibodies and immune complexes, resulting in tissue inflammation and organ damage. Besides active disease, the main cause of morbidity and mortality in SLE patients is
K Kim et al.
Tuberculosis (Edinburgh, Scotland), 94(5), 494-501 (2014-08-07)
Previous studies suggest that control of Mycobacterium tuberculosis infection is compromised by the activity of regulatory T cells, including those that express CD39, an ectonucleotidase with immunosuppressive properties. Here, we examine the role of CD39 on CD4+ T cells reacting
Chris P Verschoor et al.
Infection and immunity, 82(11), 4824-4833 (2014-08-27)
Pneumonia caused by Streptococcus pneumoniae is a major cause of death and an economic burden worldwide. S. pneumoniae is an intermittent colonizer of the human upper respiratory tract, and the ability to control asymptomatic colonization determines the likelihood of developing
Stefanie Raab et al.
Journal of immunology (Baltimore, Md. : 1950), 193(8), 4261-4272 (2014-09-14)
The ability of NK cells to mediate Ab-dependent cellular cytotoxicity (ADCC) largely contributes to the clinical success of antitumor Abs, including trastuzumab, which is approved for the treatment of breast cancer with HER2/neu overexpression. Notably, only ∼25% of breast cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service