RAB0458
Human Thyroglobulin ELISA Kit
for serum, plasma, cell culture supernatants and urine
Synonym(s):
TGF-beta-1, Thyroglobulin
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.77
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type urine
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 220 pg/mL
standard curve range: 0.205-50 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TG(7038)
General description
The Human Thyroglobulin ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human Thyroglobulin in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human Thyroglobulin
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human Thyroglobulin ELISA Kit has been used to measure the secreted thyroglobulin (TG) level in primary human thyrocytes.
Biochem/physiol Actions
Thyroglobulin (TG) is a prothyroid hormone. It plays a vital role in production of L-thyroxine (tetraiodothyronine, T4) and L-triiodothyronine (T3) hormones in thyroid gland. Tyrosyls of TG acts as substrate for iodination and hormone formation. Serum thyroglobulin level is used as marker of metastases in differentiated thyroid carcinomas. Mutation in the gene is associated with the development of congenital goitrous hypothyroidism and non-endemic simple goitre.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yuwei Wang et al.
Ecotoxicology and environmental safety, 174, 224-235 (2019-03-08)
In recent years, decabromodiphenyl ethane (DBDPE), a new alternative flame retardant to the decabrominated diphenyl ethers (BDE-209), is widely used in a variety of products. Previous studies have indicated that DBDPE, like BDE-209, could disrupt thyroid function. However, compared with
Sarah J Morgan et al.
Thyroid : official journal of the American Thyroid Association, 26(12), 1794-1803 (2016-10-19)
Major regulation of thyroid gland function is mediated by thyrotropin (TSH) activating the TSH receptor (TSHR) and inducing upregulation of genes involved in thyroid hormone synthesis. Evidence suggests that the insulin-like growth factor 1 (IGF-1) receptor (IGF-1R) may play a
Thyroglobulin gene point mutation associated with non-endemic simple goitre.
Corral J, et al.
Lancet, 341(8843), 462-464 (1993)
Role of extracellular molecular chaperones in the folding of oxidized proteins refolding of colloidal thyroglobulin by protein disulfide isomerase and immunoglobulin heavy chain-binding protein.
Delom F, et al.
The Journal of Biological Chemistry, 276(24), 21337-21342 (2001)
A nonsense mutation causes human hereditary congenital goiter with preferential production of a 171-nucleotide-deleted thyroglobulin ribonucleic acid messenger.
Targovnik H M, et al.
The Journal of Clinical Endocrinology and Metabolism, 77(1), 210-215 (1993)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service