Skip to Content
Merck
All Photos(2)

Key Documents

RAB0247

Sigma-Aldrich

Rat IL-10 ELISA Kit

for cell and tissue lysates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type tissue lysate
sample type cell lysate

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 8.23-6000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... Il10(25325)

General description

The Rat IL-10 ELSA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of Rat IL-10 cell lysate and tissue lysate.

Immunogen

Recombinant Rat IL-10

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABLYSIS12X Cell Lysis Buffer (Item J)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ian A Parish et al.
The Journal of clinical investigation, 124(8), 3455-3468 (2014-07-09)
During the course of many chronic viral infections, the antiviral T cell response becomes attenuated through a process that is regulated in part by the host. While elevated expression of the immunosuppressive cytokine IL-10 is involved in the suppression of
B Ahmed et al.
Vaccine, 32(31), 3909-3916 (2014-06-01)
Enterohemorrhagic Escherichia coli (EHEC) have been responsible for several outbreaks of hemolytic-uremic syndrome (HUS) worldwide. HUS is the most common cause of acute renal failure in children and results in fatalities as high as 50% in the elderly. Currently, neither
Bjarki Johannesson et al.
Disease models & mechanisms, 7(8), 977-985 (2014-07-25)
Allergic contact dermatitis (ACD) is triggered by an aberrant hyperinflammatory immune response to innocuous chemical compounds and ranks as the world's most prevalent occupational skin condition. Although a variety of immune effector cells are activated during ACD, regulatory T (Treg)
Yinan Deng et al.
Stem cells and development, 23(17), 2080-2092 (2014-04-16)
The mechanisms responsible for the inhibitory effects of mesenchymal stem cells (MSCs) on dendritic cells (DCs) are still poorly understood. Our investigation of the potential signaling pathways revealed for the first time that human umbilical-cord-derived MSCs (UC-MSCs) instruct DCs to
Yang Yu et al.
International immunopharmacology, 85, 106585-106585 (2020-05-25)
Sepsis-associated encephalopathy (SAE) is a cognitive impairment caused by sepsis and is related to increased morbidity and mortality. Damage to the blood-brain barrier (BBB) has been proved to be one of the important causes of SAE. Molecular hydrogen (H2) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service