Skip to Content
Merck
All Photos(2)

Key Documents

RAB0245

Sigma-Aldrich

Mouse IL-10 ELISA Kit

for serum, plasma and cell culture supernatant

Synonym(s):

IL-10, Interleukin-10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 45 pg/mL
standard curve range: 78.13-5000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Il10(16153)

General description

The Mouse IL-10 (Interleukin-10) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of IL-10 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse IL-10

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Biochem/physiol Actions

IL-10 (interleukin 10) is an anti-inflammatory cytokine.[1] In mouse, IL-10 produced by the CD4+ Th2 (T helper cell 2) subset, inhibits the expression of cytokines specifically IFN-3 (interferon), by Thl (T helper cell 1) clones. mIL-10 has pleiotropic property and it interacts with various cell types such as thymocytes, cytotoxic T cells, mast cells, B cells, and macrophages.[2] Polymorphisms in the IL-10 gene are associated with higher risk of preeclampsia, ankylosing spondylitis, nasopharyngeal carcinoma and basal cell carcinoma.[3][4][5][6] B- cell specific IL-10 might have a suppressive role in systemic lupus erythematosus (SLE).[7]

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADDELISA 5X Assay/Sample Diluent Buffer D (Item K)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IL-10 Production Is Critical for Sustaining the Expansion of CD5+ B and NKT Cells and Restraining Autoantibody Production in Congenic Lupus-Prone Mice.
Baglaenko Y
PLoS ONE, 11(3), e0150515-e0150515 (2016)
Investigations into the association between polymorphisms in the interleukin-10 gene and risk of early-onset preeclampsia.
Liu QY
Genetics and molecular research : GMR, 14(4), 19323-19328 (2015)
Clinical significance of IL-2 and IL-10 gene polymorphisms and serum levels in patients with basal-cell carcinoma.
Sobjanek M
Biomarkers in Medicine, 10(2), 185-195 (2016)
Interleukin 10(IL-10) inhibits cytokine synthesis by human monocytes: an autoregulatory role of IL-10 produced by monocytes.
de Waal Malefyt R
The Journal of Experimental Medicine, 174(5), 1209-1220 (1991)
Interleukin-10 polymorphisms and nasopharyngeal carcinoma risk: a meta-analysis.
Yu YF
Genetics and molecular research : GMR, 14(4), 18945-18957 (2015)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service