Skip to Content
Merck
All Photos(1)

Key Documents

R5278

Sigma-Aldrich

Anti-RAGE (Receptor for Advanced Glycation Endproducts) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, rat

technique(s)

immunohistochemistry: 1-2 μg/mL using transgenic mouse retina
western blot: 1 μg/mL using using mouse lung extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AGER(177)
mouse ... Ager(11596)
rat ... Ager(81722)

General description

Receptor for advanced glycation end products (RAGE), also known as advanced glycosylation end-product specific receptor (AGER) is encoded by the gene mapped to human chromosome 6p21. The encoded protein is a member of the immunoglobulin superfamily of cell surface molecules.

Immunogen

synthetic peptide corresponding to amino acid residues 362-380 from rat RAGE. This sequence is 100% conserved in mouse and 70% conserved in human.

Application

Anti-RAGE (Receptor for Advanced Glycation Endproducts) antibody produced in rabbit has been used in western blot and flow cytometry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The soluble form of receptor for advanced glycation end products (RAGE) has a function in the Wnt pathway. RAGE has a role in the development of inflammatory and cellular damage, as well as cellular toxicity. It has been associated with diabetic atherosclerosis. AGER/RAGE also modulates cell proliferation, differentiation and apoptosis.{93

Physical form

Solution in phosphate buffered saline containing 1.0 mg/mL bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of Polymorphisms of the Receptor for Advanced Glycation Endproducts Gene with Schizophrenia in a Han Chinese Population
Fu J, et al.
BioMed Research International (2017)
Is there a role for high mobility group box 1 and the receptor for advanced glycation end products in the genesis of long-term cognitive impairment in sepsis survivors?
Felipe Dal-Pizzol et al.
Molecular medicine (Cambridge, Mass.), 18, 1357-1358 (2012-11-02)
Association of RAGE gene Gly82Ser polymorphism with coronary artery disease and ischemic stroke
Ma W Q, et al.
Medicine, 95(49) (2016)
Susmita Barman et al.
Chemico-biological interactions, 307, 37-50 (2019-04-23)
Zinc depletion during diabetes postulates a role for zinc nutrition in the management of associated complications. The present study evaluated zinc supplementation for countering the compromised intestinal integrity through moderation of oxidative stress and suppression of stress-stimulated inflammatory proliferation in
Hyperglycemia induces oxidative damage in SW872 cells.
Boyer F, et al.
Archives of Medical Research, 1(2), 66-78 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service