Skip to Content
Merck

R1908

Remifentanil hydrochloride

≥97% (HPLC), Mu opioid receptor agonist, powder

Synonym(s):

3-[methoxycarbonyl-4-[(1-oxopropyl)phenylamino]1-piperidine]propanoic acid methyl ester hydrochloride, GI-87084B, Remifentanyl, Ultiva

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H28N2O5·HCl
CAS Number:
Molecular Weight:
412.91
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥97% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Remifentanil hydrochloride, ≥97% (HPLC), powder

Quality Segment

assay

≥97% (HPLC)

form

powder

drug control

USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

storage condition

desiccated

color

white to off-white

solubility

H2O: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

O=C(CC)N(C1(CCN(CCC(OC)=O)CC1)C(OC)=O)C2=CC=CC=C2.[H]Cl

InChI

1S/C20H28N2O5.ClH/c1-4-17(23)22(16-8-6-5-7-9-16)20(19(25)27-3)11-14-21(15-12-20)13-10-18(24)26-2;/h5-9H,4,10-15H2,1-3H3;1H

InChI key

WFBMIPUMYUHANP-UHFFFAOYSA-N

Gene Information

human ... OPRM1(4988)

Biochem/physiol Actions

Remifentanil hydrochloride is a mu opioid receptor agonist, anesthetic, and analgesic compound.
Remifentanil hydrochloride is a mu opioid receptor agonist, anesthetic, and analgesic compound. Remifentanil was developed as an ultra-short-acting mu opioid receptor agonist with improved pharmacodynamic properties.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Still not finding the right product?

Explore all of our products under Remifentanil hydrochloride


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT SE 2

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library