Skip to Content
Merck
All Photos(1)

Key Documents

R1781

Sigma-Aldrich

Ramoplanin

Synonym(s):

A 16686, Antibiotic A 16686

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

Assay

≥75% (as ramoplanin A2)

form

powder

solubility

H2O: soluble 10 mg/mL

antibiotic activity spectrum

Gram-positive bacteria

Mode of action

cell wall synthesis | interferes

shipped in

dry ice

storage temp.

−20°C

SMILES string

ClC1C(CCC(C1)C(NC(=O)[C@H](NC(=O)[C@@H](NC(=O)CNC(=O)C(NC(=O)[C@@H](NC(=O)C(NC(=O)[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](NC(=O)[C@H](NC(=O)C(NC(=O)[C@@H](NC(=O)\C=C/C=C\CCC)CC(=O)N)CC(=O)N)C7CCC(CC7)O)CCCN)[C@@H](O

InChI

1S/C106H170ClN21O30/c1-8-9-10-11-15-22-80(140)114-75(50-78(110)138)96(147)118-76(51-79(111)139)97(148)124-86(60-25-36-66(133)37-26-60)102(153)117-72(21-17-46-109)92(143)120-83(56(6)130)100(151)126-89(63-31-42-69(136)43-32-63)105(156)127-88(62-29-40-68(135)41-30-62)104(155)121-82(55(5)129)99(150)119-74(48-58-18-13-12-14-19-58)95(146)116-71(20-16-45-108)93(144)123-87(61-27-38-67(134)39-28-61)103(154)122-84(57(7)131)101(152)125-85(59-23-34-65(132)35-24-59)98(149)112-52-81(141)115-73(47-53(2)3)94(145)113-54(4)91(142)128-90(106(157)158)64-33-44-77(137)70(107)49-64/h10-15,18-19,22,53-57,59-77,82-90,129-137H,8-9,16-17,20-21,23-52,108-109H2,1-7H3,(H2,110,138)(H2,111,139)(H,112,149)(H,113,145)(H,114,140)(H,115,141)(H,116,146)(H,117,153)(H,118,147)(H,119,150)(H,120,143)(H,121,155)(H,122,154)(H,123,144)(H,124,148)(H,125,152)(H,126,151)(H,127,156)(H,128,142)(H,157,158)/b11-10-,22-15-/t54-,55+,56+,57+,59?,60?,61?,62?,63?,64?,65?,66?,67?,68?,69?,70?,71-,72-,73+,74+,75+,76?,77?,82+,83-,84+,85?,86-,87?,88-,89-,90?/m1/s1

InChI key

FSBZBQUUCNYWOK-YIOPJBSBSA-N

General description

Chemical structure: glycopeptide
Ramoplanin is a glycolipodepsipeptide antibiotic produced by Actinoplanes sp. It is a complex of structurally related molcules, with ramoplanin A2 as the primary component. It is used for in vitro susceptibility testing and to study antibiotic-resistant enterococci.

Biochem/physiol Actions

Ramoplatin is an antimicrobial which is effective against Gram-positive bacteria, including vancomycin-resistant enterococci. It acts by inhibiting bacterial cell wall synthesis, although its mechanism of action is different from that of glycopeptide-based synthesis inhibitors.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mara Brunati et al.
The Journal of antibiotics, 58(7), 473-478 (2005-09-16)
Ramoplanin is a glycolipodepsipeptide antibiotic obtained by fermentation of the Actinoplanes sp. ATCC 33076, isolated as a complex of three closely related components A1, A2 and A3, which differ in their fatty acid moiety. We have investigated the influence of
T Peláez et al.
Antimicrobial agents and chemotherapy, 49(3), 1157-1159 (2005-02-25)
We evaluated the in vitro activity of ramoplanin, an antimicrobial compound that inhibits cell wall synthesis by acting at the level of lipid intermediate formation, against Clostridium difficile. We included strains with reduced susceptibilities to vancomycin (vancomycin-intermediate [Van(i)] strains) or
C C Johnson et al.
Antimicrobial agents and chemotherapy, 36(10), 2342-2345 (1992-10-01)
Ramoplanin, a new lipoglycodepsipeptide antibiotic, was uniformly active against 65 strains of enterococci, including strains highly resistant to vancomycin, penicillin G, and gentamicin. MBCs were usually within a fourfold dilution of the MICs. In time-kill studies, ramoplanin alone demonstrated dose-dependent
Antibiotic action and peptidoglycan formation on tethered lipid bilayer membranes.
Michael J Spencelayh et al.
Angewandte Chemie (International ed. in English), 45(13), 2111-2116 (2006-02-25)
Dongwoo Shin et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(33), 11977-11979 (2004-06-04)
Ramoplanin is a potent antibiotic, first disclosed in 1984, that acts by inhibiting bacterial cell-wall biosynthesis. The original ramoplanin complex was shown to consist of a mixture of three closely related compounds, ramoplanin A1-A3, of which ramoplanin A2 is the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service