Skip to Content
Merck
All Photos(1)

Key Documents

R103

Sigma-Aldrich

Ritanserin

powder

Synonym(s):

6-[2-[4-(bis(4-Fluorophenyl)methylene]-1-piperidinyl]ethyl]-7-methyl-5H-thiazolo[3,2-a]pyrimidin-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H25F2N3OS
CAS Number:
Molecular Weight:
477.57
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

powder

color

white

solubility

methanol: >10 mg/mL
DMSO: >10.1 mg/mL
2-hydroxypropyl-β-cyclodextrin: insoluble
H2O: insoluble
dilute aqueous base: insoluble

SMILES string

CC1=C(CCN2CCC(\CC2)=C(\c3ccc(F)cc3)c4ccc(F)cc4)C(=O)N5C=CSC5=N1

InChI

1S/C27H25F2N3OS/c1-18-24(26(33)32-16-17-34-27(32)30-18)12-15-31-13-10-21(11-14-31)25(19-2-6-22(28)7-3-19)20-4-8-23(29)9-5-20/h2-9,16-17H,10-15H2,1H3

InChI key

JUQLTPCYUFPYKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ritanserin has been used in behavioral testing[1] and as a serotonin 2A receptor (5-HT2A)-receptor antagonist.[2][3]

Biochem/physiol Actions

Potent 5-HT2A serotonin receptor antagonist/inverse agonist that crosses the blood-brain barrier.
Ritanserin is a potent serotonin 2A receptor (5-HT2A) serotonin receptor antagonist/inverse agonist, which crosses the blood-brain barrier.[4] It modulates 5-HT2A receptors and has been suggested to have antiparkinsonian effect.[1] Ritanserin also impacts schizophrenia by modulating mood, cognition and negative symptoms.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Depression-like behavior in the forced swimming test in PACAP-deficient mice: amelioration by the atypical antipsychotic risperidone
Hashimoto H, et al.
Journal of Neurochemistry, 110(2), 595-602 (2009)
Does ritanserin, a potent serotonin-S2 antagonist, restore energetic functions during the night?
Janssen, PAJ
Journal of the Royal Society of Medicine, 80, 409-413 (1987)
Modulation by 5-HT2A receptors of aggressive behavior in isolated mice
Sakaue M, et al.
Japanese Journal of Pharmacology, 89(1), 89-92 (2002)
Xiaojun Jia et al.
Neuropharmacology, 62(2), 775-783 (2011-09-20)
The key role of the hypothalamic neuropeptides orexins in maintenance and promotion of arousal has been well established in normal mammalian animals, but whether orexins exert arousal effects under pathological condition such as coma was little studied. In this study
Agnieszka Pałucha-Poniewiera et al.
The Journal of pharmacology and experimental therapeutics, 334(3), 1066-1074 (2010-06-22)
Behavioral studies show that modulation of the glutamatergic system might be an efficient way to achieve antidepressant activity. Among the group III metabotropic glutamate (mGlu) receptors, the mGlu7 receptor subtype seems to be the most promising target for potential antidepressants.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service