Skip to Content
Merck
All Photos(2)

Key Documents

R0881

Sigma-Aldrich

Anti-Rabbit IgG (whole molecule) antibody produced in goat

whole antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

radioimmunoassay: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG antibodies are mainly responsible for the facilitation of humoral immune responses such as complement fixation, placental transport, and phagocytosis . Anti-rabbit IgG (whole molecule) antibody can be used in immunoelectrophoresis. Goat anti- rabbit IgG antibody reacts specifically with IgG of rabbit.
IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, and viruses.

Immunogen

rabbit IgG (whole molecule)

Application

Anti-Rabbit IgG (whole molecule) antibody produced in goat has been used in western blotting and immunofluorescent staining.
Goat anti-rabbit IgG (whole molecule) has been used in
  • radioimmunoassay
  • western blotting
  • immunofluorescent staining

Biochem/physiol Actions

IgG antibodies are mainly responsible for the facilitation of humoral immune responses such as complement fixation, placental transport, and phagocytosis ..
Maternal IgG is transferred to fetus through the placenta that is vital for immune defence of the neonate against infections.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lili Bai et al.
Cancer management and research, 13, 259-275 (2021-01-21)
To explore the role and potential mechanism of circSND1 in cervical cancer (CC). qRT-PCR was used to determine the expression of circSND1 in tumor necrosis factor-α (TNF-α)-treated HeLa cells. CircSND1 overexpression and knockdown were performed to indicate the functional role
Ningping Gong et al.
Endocrinology, 154(7), 2331-2340 (2013-05-07)
In mammals, leptin (Lep) binding proteins (LepBPs) derived from Lep receptor (LepR) gene or protein bind most of the circulating Lep, but to date, information on LepBPs in nonmammalian vertebrate classes is lacking. This study details the characterization of multiple
Cheryl S Watson et al.
Steroids, 72(2), 124-134 (2006-12-19)
Studies of the nuclear transcriptional regulatory activities of non-physiological estrogens have not explained their actions in mediating endocrine disruption in animals and humans at the low concentrations widespread in the environment. However, xenoestrogens have rarely been tested for their ability
Cheryl S Watson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 22(9), 3328-3336 (2008-06-11)
Physiological estrogens, including estrone (E(1)), estradiol (E(2)), and estriol (E(3)), fluctuate with life stage, suggesting specific roles for them in biological and disease processes. We compared their nongenomic signaling and functional actions in GH3/B6/F10 rat pituitary tumor cells. All hormones
Ken-ichi Yayou et al.
Animal science journal = Nihon chikusan Gakkaiho, 85(4), 481-493 (2013-11-23)
To examine the influence of postnatal oxytocin (OXT) on behavioral development, we examined individual variations in basal OXT and suckling-induced changes in OXT, and their relationship with emotional traits during development. Blood OXT were measured in 20 Holstein heifer calves

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service