Skip to Content
Merck
All Photos(1)

Key Documents

Q111

Sigma-Aldrich

(±)-Quinpirole dihydrochloride

powder

Synonym(s):

trans-(±)-4,4a,5,6,7,8,8a,9-Octahydro-5-propyl-1H-pyrazolo[3,4-g]quinoline dihydrochloride, LY-141,865

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H21N3 · 2HCl
CAS Number:
Molecular Weight:
292.25
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

powder

color

white

solubility

H2O: 28 mg/mL

storage temp.

−20°C

SMILES string

Cl.Cl.[H][C@]12CCCN(CCC)[C@]1([H])Cc3cn[nH]c3C2

InChI

1S/C13H21N3.2ClH/c1-2-5-16-6-3-4-10-7-12-11(8-13(10)16)9-14-15-12;;/h9-10,13H,2-8H2,1H3,(H,14,15);2*1H/t10-,13-;;/m1../s1

InChI key

GJIGRGIGKHPYTK-OWVUFADGSA-N

Gene Information

human ... DRD2(1813)

Biochem/physiol Actions

D2-like dopamine receptor agonist that possesses some activity at D3 sites.

Legal Information

Manufactured and sold with the permission of Eli Lilly and Company.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cristina Cariulo et al.
Biochemical and biophysical research communications, 521(3), 549-554 (2019-11-05)
Huntington's disease (HD) is a progressive neurodegenerative disorder caused by an expansion of a CAG triplet repeat (encoding for a polyglutamine tract) within the first exon of the huntingtin gene. Expression of the mutant huntingtin (mHTT) protein can result in
Gunnar F Kwakye et al.
Neurotoxicology, 70, 48-61 (2018-11-07)
Huntington's disease (HD) is functionally linked to environmental factors including cigarette use and dyshomeostasis in the levels of metals. Interestingly, one of the most abundant heavy metals in cigarettes is cadmium (Cd), which also accumulates in the striatum and causes
D Eilam et al.
European journal of pharmacology, 161(2-3), 151-157 (1989-02-28)
The effects of the D-2 agonist quinpirole on forward progression, and on vertical and lateral movements, were measured for 2 h in rats injected with either saline, 0.03, 0.125, 0.5 or 8 mg/kg of the drug. Results showed that the
Laura Casares et al.
Redox biology, 37, 101689-101689 (2020-08-31)
Oxidative stress and inflammation in the brain are two key hallmarks of neurodegenerative diseases (NDs) such as Alzheimer's, Parkinson's, Huntington's and multiple sclerosis. The axis NRF2-BACH1 has anti-inflammatory and anti-oxidant properties that could be exploited pharmacologically to obtain neuroprotective effects.
L J Munro et al.
Brain research, 746(1-2), 231-238 (1997-01-23)
Dopamine (DA) D2 and gamma-aminobutyric acid (GABA)A somatodendritic receptors tonically inhibit mesolimbic projection neurons in the A10 DA cell grouping of the ventral tegmentum. In the present study we determined the contribution of the ventral tegmental area (VTA) to the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service