Skip to Content
Merck
All Photos(1)

Key Documents

PZ0160

Sigma-Aldrich

UK-5099

≥98% (HPLC), powder, mitochondrial pyruvate carrier inhibitor

Synonym(s):

2-Cyano-3-(1-phenyl-1H-indol-3-yl)-2-propenoic acid, PF-1005023

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12N2O2
CAS Number:
Molecular Weight:
288.30
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Product Name

UK-5099, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

color

yellow to tan

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)\C(=C\c1cn(-c2ccccc2)c3ccccc13)C#N

InChI

1S/C18H12N2O2/c19-11-13(18(21)22)10-14-12-20(15-6-2-1-3-7-15)17-9-5-4-8-16(14)17/h1-10,12H,(H,21,22)/b13-10+

InChI key

BIZNHCWFGNKBBZ-JLHYYAGUSA-N

Related Categories

Application

UK-5099 has been used:
  • as a mitochondrial pyruvate blocker to reduce pyruvate transportation into mitochondria in Roswell park memorial institute (RPMI) 1640 medium for prostatic cancer cell line culture
  • in dimethyl sulfoxide (DMSO) stock, to study the effect of inhibiting pyruvate transport into mitochondria on pro-inflammatory responses in lipopolysaccharide activated macrophages
  • in topical treatment in order to study its effect on hair cycle induction in experimental mice

Biochem/physiol Actions

UK-5099 induces lactate generation.
UK-5099 is a potent inhibitor of the mitochondrial pyruvate transporter.
UK-5099 is a potent inhibitor of the mitochondrial pyruvate transporter. In rat heart mitochondria, UK-5099 inhibits pyruvate-dependent O2 consumption with an IC50 of 50 nM.

Features and Benefits

This compound is a featured product for Apoptosis research. Click here to discover more featured Apoptosis products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A P Halestrap et al.
The Biochemical journal, 223(3), 677-685 (1984-11-01)
The inhibitor of mitochondrial pyruvate transport alpha-cyano-beta-(1-phenylindol-3-yl)-acrylate was used to inhibit progressively pyruvate carboxylation by liver mitochondria from control and glucagon-treated rats. The data showed that, contrary to our previous conclusions [Halestrap (1978) Biochem. J. 172, 389-398], pyruvate transport could
Oleic acid stimulates glucagon-like peptide-1 release from enteroendocrine cells by modulating cell respiration and glycolysis
Clara R, et al.
Metabolism and Disease, 65(3), 8-17 (2016)
M O Proudlove et al.
The Biochemical journal, 247(2), 441-447 (1987-10-15)
1. Mitochondria isolated from the thermogenic spadices of Arum maculatum and Sauromatum guttatum plants oxidized external NADH, succinate, citrate, malate, 2-oxoglutarate and pyruvate without the need to add exogenous cofactors. 2. Oxidation of substrates was virtually all via the alternative
M S Shearman et al.
The Biochemical journal, 223(3), 673-676 (1984-11-01)
alpha-Cyano-beta-(1-phenylindol-3-yl)acrylate inhibited pyruvate transport into both liver and heart mitochondria approximately linearly with respect to its concentration until 65% inhibition was achieved. The extent of inhibition was dependent on the mitochondrial protein concentration. By extrapolation of plots of inhibition versus
Lactate dehydrogenase activity drives hair follicle stem cell activation
Flores A, et al.
Nature Cell Biology, 19(9), 1017-1017 (2017)

Articles

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

Cell cycle phases (G1, S, G2, M) regulate cell growth, DNA replication, and division in proliferating cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service