Skip to Content
Merck
All Photos(3)

Key Documents

PRS2397

Sigma-Aldrich

Anti-BCMA antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Tumor necrosis factor receptor superfamily, member 17

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 20 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF17(608)

General description

B-cell maturation antigen (BCMA), commonly known as TNFRSF17 (tumor necrosis factor receptor superfamily member 17), is an N-glycosylated membrane glycoprotein. It is predominantly expressed on the surface of plasma cells, and is also expressed on certain cancers such as, glioblastoma, chronic lymphocytic leukemia, Hodgkin′s lymphoma and multiple myeloma.

Immunogen

a synthetic peptide mapping at the carboxy-terminus of human BCMA.

Biochem/physiol Actions

TNFRSF17 (tumor necrosis factor receptor superfamily member 17) is essential for the maintenance of the survival of long-lived plasma cells in bone marrow. Its expression in peripheral blood B-cells is induced by cytokines. It gets activated by a proliferation-inducing ligand (APRIL) or B-cell–activating factor (BAFF), and gets trimerized to activate MAPK (mitogen activated kinases) and anti-apoptotic proteins. It is an appropriate target for T-cells expressing chimeric antigen receptor (CAR), and is a promising candidate for adoptive T-cell therapy of multiple myeloma. This protein is directly cleaved and shed by γ-secretase enzyme, and this is linked with level of plasma cells in bone marrow. Thus, shedding of TNFRSF17 is a candidate biomarker for B-cell involvement in autoimmune diseases. This protein is thought to be a major contributor of spinal cord-injury induced autoimmunity.

Linkage

The action of this antibody can be blocked using blocking peptide SBP2397.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han-Wen Huang et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(27), 10928-10933 (2013-06-19)
Glycosylation, an important posttranslational modification process, can modulate the structure and function of proteins, but its effect on the properties of plasma cells is largely unknown. In this study, we identified a panel of glycoproteins by click reaction with alkynyl
Jonah W Saltzman et al.
Journal of neurotrauma, 30(6), 434-440 (2012-10-24)
Autoimmunity is thought to contribute to poor neurological outcomes after spinal cord injury (SCI). There are few mechanism-based therapies, however, designed to reduce tissue damage and neurotoxicity after SCI because the molecular and cellular bases for SCI-induced autoimmunity are not
Robert O Carpenter et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 19(8), 2048-2060 (2013-01-25)
Multiple myeloma is a usually incurable malignancy of plasma cells. New therapies are urgently needed for multiple myeloma. Adoptive transfer of chimeric antigen receptor (CAR)-expressing T cells is a promising new therapy for hematologic malignancies, but an ideal target antigen
Sarah A Laurent et al.
Nature communications, 6, 7333-7333 (2015-06-13)
Survival of plasma cells is regulated by B-cell maturation antigen (BCMA), a membrane-bound receptor activated by its agonist ligands BAFF and APRIL. Here we report that γ-secretase directly cleaves BCMA, without prior truncation by another protease. This direct shedding is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service