Skip to Content
Merck
All Photos(1)

Key Documents

PLA0302

Sigma-Aldrich

Anti-GAPDH Antibody

goat polyclonal

Synonym(s):

G3PD, GAPD, GAPDH, HEL-S-162eP, aging-associated gene 9 protein, epididymis secretory sperm binding protein Li 162eP, peptidyl-cysteine S-nitrosylase GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

Product Name

Goat anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.

biological source

goat

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:500-1:2,000
western blot: 1:500- 1:2,500

accession no.

NP_002037.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

goat ... GAPDH(2597)

Immunogen

The epitope recognized by PLA0302 maps to a region between residue 175 and 225 of human Glyceraldehyde-3-Phosphate Dehydrogenase using the numbering given in entry NP_002037.2 (GeneID 2597).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

In the process of carbohydrate metabolism, GAPDH (glyceraldehydes-3-phosphate dehydrogenase) catalyzes the oxidative phosphorylation of glyceraldehydes-3-phosphate to 1,3-diphosphoglycerate in the presence of nicotinamide adenine dinucleotide (NAD) and inorganic phosphate. In addition to its role in the oxidation of glucose, GAPDH has been shown to be involved in other cellular processes such as vesicle transport, DNA repair, DNA replication, cytoskeletal modulation, and tRNA export.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaolei Lian et al.
Acta neuropathologica communications, 9(1), 88-88 (2021-05-19)
Diffuse intrinsic pontine glioma (DIPG), a rare pediatric brain tumor, afflicts approximately 350 new patients each year in the United States. DIPG is noted for its lethality, as fewer than 1% of patients survive to five years. Multiple clinical trials
Sarah McCollum et al.
Pharmaceuticals (Basel, Switzerland), 15(6) (2022-06-25)
Synovial sarcoma (SS) is a pediatric muscle cancer that primarily affects adolescents and young adults and has few treatment options. Complicating the treatment of synovial sarcoma is the low mutational burden of SS. Inflammatory pathways have been identified as being
Qiyue Ding et al.
International journal of molecular sciences, 22(4) (2021-02-12)
Alzheimer's disease is a chronic neurodegenerative disorder and represents the main cause of dementia globally. Currently, the world is suffering from the coronavirus disease 2019 (COVID-19) pandemic, caused by severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2), a virus that uses

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service