Skip to Content
Merck
All Photos(4)

Key Documents

PLA0227

Sigma-Aldrich

Rabbit anti-BRD4 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

CAP, HUNK1, HUNKI, MCAP, Protein HUNK1, bromodomain-containing 4, chromosome-associated protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry: 1:1,000-1:5,000
immunoprecipitation (IP): 2-5 μg/mg
western blot: 1:2,000-1:10,000

accession no.

NP_490597.1

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... BRD4(23476)

General description

Bromodomain-containing protein 4 (BRD4) belongs to the bromodomain and extra-terminal (BET) family. The protein has two bromodomains (BD1 and BD2) connected by a long interdomain linker.

Immunogen

The epitope recognized by PLA0227 maps to a region between residue 1312 and 1362 of human bromodomain-containing protein 4 using the numbering given in entry NP_490597.1 (GeneID 23476).

Biochem/physiol Actions

Bromodomain-containing protein 4 (BRD4) works as a reader of epigenetic marks and is involved in chromatin remodeling and transcriptional regulation. Therefore, it is a target for oncology, inflammation and cardiovascular disease. This gene is upregulated in hepatocellular carcinoma and is associated with poor prognosis. BRD4 is also associated with melanoma, hepatocellular carcinoma, multiple myeloma, Burkitt′s lymphoma, acute myeloid leukemia and breast cancer. BRD4 connects cell cycle and transcription, works as a scaffold for transcription factors and bookmarks active genes during mitosis. It interacts with acetylated histone-3 and histone-4 tails and helps in maintaining chromatin architecture. Also, it regulates transcription elongation via phosphorylation of RNA polymerase II.

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

Bromodomain-containing protein 4 (BRD4) contains 2 bromo-domains. Bromo-domains are found in chromatin-associating proteins and can interact specifically with acetylated lysine. BRD4 is described as a chromatin adaptor and has been shown to bind acetylated chromatin. It was originally identified as MCAP (mitotic chromosome associated protein) due to its association with mitotic chromosomes. BRD4 also associates with several transcription complexes such as the Mediator complexes and P-TEFb complexes and is considered to function as a transcriptional co-factor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

BRD4 promotes tumor growth and epithelial-mesenchymal transition in hepatocellular carcinoma.
Zhang P, et al.
International Journal of Immunopathology and Pharmacology, 28(1), 36-44 (2015)
Segmental Isotope Labelling of an Individual Bromodomain of a Tandem Domain BRD4 Using Sortase A.
Williams F P, et al.
PLoS ONE, 11(4), e0154607-e0154607 (2016)
High level of BRD4 promotes non-small cell lung cancer progression.
Liao Y F, et al.
Oncotarget, 7(8), 9491-9491 (2016)
Shun-Hong Luo et al.
Cell death & disease, 14(11), 760-760 (2023-11-23)
Lipid metabolism is the key to ferroptosis susceptibility. However, little is known about the underlying mechanisms in osteosarcoma cells. Functional restriction of bromodomain-containing protein 4 (BRD4) reduced the susceptibility to erastin-induced ferroptosis of osteosarcoma cells both in vitro and in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service