Skip to Content
Merck
All Photos(2)

Key Documents

PLA0125

Sigma-Aldrich

Anti-GAPDH Antibody

rabbit polyclonal

Synonym(s):

G3PD, GAPD, GAPDH, HEL-S-162eP, aging-associated gene 9 protein, epididymis secretory sperm binding protein Li 162eP, peptidyl-cysteine S-nitrosylase GAPDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Rabbit anti-GAPDH Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:100-1:500
western blot: 1:10,000-1:25,000

accession no.

NP_002037.2

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... GAPDH(2597)

Immunogen

The epitope recognized by PLA0125 maps to a region between residues 250 and 300 of human Glyceraldehyde-3-Phosphate Dehydrogenase using the numbering given in entry NP_002037.2 (GeneID 2597).

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

In the process of carbohydrate metabolism, GAPDH (glyceraldehydes-3-phosphate dehydrogenase) catalyzes the oxidative phosphorylation of glyceraldehydes-3-phosphate to 1,3-diphosphoglycerate in the presence of nicotinamide adenine dinucleotide (NAD) and inorganic phosphate. In addition to its role in the oxidation of glucose, GAPDH has been shown to be involved in other cellular processes such as vesicle transport, DNA repair, DNA replication, cytoskeletal modulation, and tRNA export.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changlong Li et al.
Oncology letters, 15(6), 9673-9680 (2018-06-22)
Multidrug resistance (MDR) is a major obstacle to cancer chemotherapy efficacy. In the present study, 6-O-angeloylplenolin repressed the overexpression of ATP binding cassette subfamily B member 1 (MDR1) and increasing the intracellular concentration of anticancer drugs. A reduction in P-glycoprotein
Renlong Li et al.
Obesity (Silver Spring, Md.), 25(3), 561-571 (2017-02-02)
1,25(OH)2 D3 has been reported to attenuate liver steatosis; however, its exact mechanism of action remains poorly understood. This study aimed to determine whether 1,25(OH)2 D3 can attenuate hepatic steatosis by inducing autophagy. Male C57BL/6 mice fed a high-fat diet
Remi Sonneville et al.
eLife, 8 (2019-09-24)
The faithful segregation of eukaryotic chromosomes in mitosis requires that the genome be duplicated completely prior to anaphase. However, cells with large genomes sometimes fail to complete replication during interphase and instead enter mitosis with regions of incompletely replicated DNA.
Yoshihiro Ishikawa et al.
The Journal of biological chemistry, 296, 100453-100453 (2021-02-26)
Collagen is the most abundant protein in humans. It has a characteristic triple-helix structure and is heavily posttranslationally modified. The complex biosynthesis of collagen involves processing by many enzymes and chaperones in the rough endoplasmic reticulum. Lysyl hydroxylase 1 (LH1)
Chunyang Wang et al.
Cancer science, 106(4), 375-382 (2015-02-04)
Prostate cancer (PCa) prevails as the most commonly diagnosed malignancy in men and the third leading cause of cancer-related deaths in developed countries. One of the distinct characteristics of prostate cancer is overexpression of the small ubiquitin-like modifier (SUMO)-specific protease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service