Skip to Content
Merck
All Photos(1)

Key Documents

P9764

Sigma-Aldrich

Polydeoxyadenylic acid • Polythymidylic acid sodium salt

double-stranded homopolymer

Synonym(s):

Poly(dA) • Poly(dT)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.51

biological source

synthetic (organic)

Quality Level

form

powder

storage temp.

−20°C

SMILES string

[P](=O)(OCC4OC(CC4O)N5C=C(C(=O)NC5=O)C)(O)O.[P](=O)(OCC1OC(CC1O)[n]2c3ncnc(c3nc2)N)(O)O

InChI

1S/C10H14N5O6P.C10H15N2O8P/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(21-7)2-20-22(17,18)19;1-5-3-12(10(15)11-9(5)14)8-2-6(13)7(20-8)4-19-21(16,17)18/h3-7,16H,1-2H2,(H2,11,12,13)(H2,17,18,19);3,6-8,13H,2,4H2,1H3,(H,11,14,15)(H2,16,17,18)

InChI key

KKVQVIFXABFJCS-UHFFFAOYSA-N

Application

Polydeoxyadenylic acid • Polythymidylic acid (Poly(dA) • Poly(dT)) is a double-stranded homopolymer use as a DNA model for conformational studies of DNA structure dynamics and drug interaction.

Other Notes

Double-stranded homopolymer

Unit Definition

One unit will yield an A260 of 1.0 in 1.0 ml 20 mM sodium phosphate/100 mM NaCl, pH 7.0

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Cao et al.
Journal of biomolecular structure & dynamics, 18(6), 844-856 (2001-07-11)
The DNA binding behavior of a tricationic cyanine dye (DiSC3+(5)) was studied using the [Poly(dA-dT)]2, [Poly(dI-dC)]2 and Poly(dA) x Poly(dT) duplex sequences and the Poly(dA) x 2Poly(dT) triplex. Optical spectroscopy and viscometry results indicate that the dye binds to the
U Sehlstedt et al.
Journal of molecular biology, 278(1), 31-56 (1998-05-26)
The binding of an antiviral quinoxaline derivative, 2,3-dimethyl- 6 - (dimethylaminoethyl) - 9 - hydroxy - 6H - indolo - [2,3 - b]quinoxaline (9-OH-B220), to synthetic double and triple helical DNA (poly(dA).poly(dT) and poly(dA).2poly(dT)) and RNA (poly(rA). poly(rU) and poly
K M Kosikov et al.
Journal of molecular biology, 289(5), 1301-1326 (1999-06-22)
Computer-simulated elongation and compression of A - and B -DNA structures beyond the range of thermal fluctuations provide new insights into high energy "activated" forms of DNA implicated in biochemical processes, such as recombination and transcription. All-atom potential energy studies
M Thomas et al.
Journal of biomolecular structure & dynamics, 18(6), 858-871 (2001-07-11)
The binding of three analogues of the minor-groove binding antiviral antibiotic distamycin (Dst) with double-stranded (ds)-DNA were monitored using ds-DNA melting temperature (Tm) measurements, ethidium bromide (EtBr) displacement assay, footprinting analysis and induced circular dichroism (ICD). These compounds contained 3-5
Hongjuan Xi et al.
Biochemistry, 50(42), 9088-9113 (2011-08-26)
Recognition of nucleic acids is important for our understanding of nucleic acid structure as well as for our understanding of nucleic acid-protein interactions. In addition to the direct readout mechanisms of nucleic acids such as H-bonding, shape recognition of nucleic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service