Skip to Content
Merck
All Photos(1)

Key Documents

P9523

Sigma-Aldrich

Plasma

from human

Synonym(s):

Citrated plasma

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.71

biological source

human

Quality Level

form

powder

contains

4% trisodium citrate as anticoagulant

impurities

HIV, hepatitis B and hepatitis C, none detected

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Plasma is the liquid part of the blood and lymphatic fluid, which makes up about half of its volume. Plasma is devoid of cells and, unlike serum, has not clotted. It is prepared from whole blood that is collected with anticoagulants (9:1) and centrifuged to remove cells and cellular debris. Some of the more common proteins found in plasma are albumin and prealbumin, α1-acid glycoprotein, transferrin, lipoproteins (HDL, LDL, and VLDL), immunoglobulins, complement proteins, and coagulation proteins (thrombin, plasminogin, and fibrinogen). Citrated plasma contains citrate (4% trisodium citrate) as an anticoagulant.

Plasma contains a variety of proteins with diverse functions. The primary functions of the plasma proteins include the maintenance of colloid osmotic pressure, pH, and electrolyte balance; the transport of metal ions, fatty acids, steroids, hormones, and drugs to various organs of the body; use as a source for amino acids for tissue nourishment; hemostasis and the prevention of thrombosis; the regulation of cellular activity and function through hormone signaling; and defense against invasion through the actions of antibodies and complement components.

Other Notes

This product is prepared from pooled human blood. It contains 4% trisodium citrate as an anticoagulant. It is tested for clotting, which indicates that the clotting factors in the product are active. However, it is not analyzed to determine whether other enzymes present are native or denatured.

Preparation Note

Whole blood collected by standard industry method using citrate based anticoagulant. Whole blood is pooled and then centrifuged. The resulting plasma is 0.45 micron filtered and lyophilized from the indicated volumes.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shu Hui Neo et al.
Scientific reports, 7(1), 16686-16686 (2017-12-02)
The preservation of nucleic acids from clinical samples is critical to facilitate accurate molecular diagnosis. The use of a paper matrix, Flinders Technology Associates (FTA) Elute cards, to archive DNA and viral RNA is well-documented. However, the feasibility of FTA
Sofia G Meirinho et al.
Analytica chimica acta, 987, 25-37 (2017-09-17)
A DNA aptamer with affinity and specificity for human osteopontin (OPN), a potential breast cancer biomarker, was selected using the SELEX process, considering its homology rate and the stability of its secondary structures. This aptamer exhibited a satisfactory affinity towards
Noor Al-Saden et al.
Molecular pharmaceutics, 15(8), 3383-3393 (2018-06-30)
Our aim was to synthesize 89Zr-labeled trastuzumab-emtansine (89Zr-DFO-T-DM1) to probe the delivery of trastuzumab-emtansine (T-DM1) to HER2-positive breast cancer (BC) by positron emission tomography (PET). We further aimed to compare the tumor and normal tissue uptake of 89Zr-DFO-T-DM1 with 89Zr-DFO-trastuzumab.
F Vida Zohoori et al.
Caries research, 53(3), 275-283 (2018-10-09)
The aim was to compare potential methods for fluoride analysis in microlitre-volume plasma samples containing nano-gram amounts of fluoride. Methods: A group of 4 laboratories analysed a set of standardised biological samples as well as plasma to determine fluoride concentration
Karin Löw et al.
PloS one, 15(5), e0231163-e0231163 (2020-05-06)
Examination of genetic polymorphisms in outbred wild-living species provides insights into the evolution of complex systems. In higher vertebrates, the proopiomelanocortin (POMC) precursor gives rise to α-, β-, and γ-melanocyte-stimulating hormones (MSH), which are involved in numerous physiological aspects. Genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service