Skip to Content
Merck
All Photos(1)

Key Documents

P6777

Sigma-Aldrich

Phenelzine sulfate salt

Synonym(s):

Phenelzine hydrogen sulphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H12N2 · H2O4S
CAS Number:
Molecular Weight:
234.27
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥98% (iodine, titration)

form

powder

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

NNCCC1=CC=CC=C1.OS(=O)(O)=O

InChI

1S/C8H12N2.H2O4S/c9-10-7-6-8-4-2-1-3-5-8;1-5(2,3)4/h1-5,10H,6-7,9H2;(H2,1,2,3,4)

InChI key

RXBKMJIPNDOHFR-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Non-selective MAO-A/B inhibitor.

Features and Benefits

This compound is featured on the Dopamine and Norepinephrine Metabolism page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of riluzole augmentation in a patient with treatment-resistant obsessive-compulsive disorder, taking two other glutaminergic agents.
Nahla Mahgoub et al.
The Journal of neuropsychiatry and clinical neurosciences, 23(2), E24-E25 (2011-06-17)
Hwan-Suck Chung et al.
Neurochemical research, 37(10), 2117-2124 (2012-07-06)
Phenelzine is a potent monoamine oxidase inhibitor that is used in patients with depression. It is also well known that nitric oxide (NO) synthase inhibitors show preclinical antidepressant-like properties, which suggests that NO is involved in the pathogenesis of depression.
Mee-Sook Song et al.
Journal of neurochemistry, 114(5), 1405-1413 (2010-06-19)
Reactive aldehydes have been implicated in the etiology of several neurological and psychiatric disorders, and there is considerable interest in drugs to counteract the actions of these aldehydes. Increased formaldehyde (FA) and up-regulation of semicarbazide-sensitive amine oxidase, which forms FA
Travis Musgrave et al.
Brain, behavior, and immunity, 25(8), 1677-1688 (2011-07-05)
Multiple sclerosis (MS) and the animal model, experimental autoimmune encephalomyelitis (EAE), are both accompanied by motor and non-motor symptoms. Pathological changes in the activities of key neurotransmitters likely underlie many of these symptoms. We have previously described disturbances in the
Marco De Giovanni et al.
Science immunology, 9(93), eadj7363-eadj7363 (2024-03-01)
Peyer's patches (PPs) are lymphoid structures situated adjacent to the intestinal epithelium that support B cell responses that give rise to many intestinal IgA-secreting cells. Induction of isotype switching to IgA in PPs requires interactions between B cells and TGFβ-activating

Articles

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Serotonin is stored in cells and metabolized by MAO, influencing CNS, GI, and platelet functions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service