Skip to Content
Merck
All Photos(2)

Key Documents

P5869

Sigma-Aldrich

Phosphatase substrate

chromogenic, powder

Synonym(s):

4-Nitrophenyl phosphate disodium salt hexahydrate, pNPP disodium salt hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

product name

Phosphatase substrate, 100 mg capsules

Quality Level

form

powder

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Chromogenic substrate for use with alkaline and acid phosphatase ELISA procedures. Produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 N NaOH and read at 405 nm.

Other Notes

Substrate for the determination of acid and alkaline phosphatases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jungju Kim et al.
Tissue engineering and regenerative medicine, 18(3), 387-397 (2021-01-09)
Three-dimensional (3D) in vitro cultures recapitulate the physiological microenvironment and exhibit high concordance with in vivo conditions. Improving co-culture models with different kind of cell types cultured on a 3D scaffold can closely mimic the in vivo environment. In this
E Swilks et al.
Australian veterinary journal, 95(6), 211-216 (2017-05-31)
To examine the prevalence of Theileria orientalis in the Gloucester area of New South Wales and its effect on individual animals. Blood samples (EDTA and clotted blood) were collected from a total of 55 calves and their dams from 6
Miriam Filippi et al.
Biomaterials, 223, 119468-119468 (2019-09-11)
Exposure of cells to externally applied magnetic fields or to scaffolding materials with intrinsic magnetic properties (magnetic actuation) can regulate several biological responses. Here, we generated novel magnetized nanocomposite hydrogels by incorporation of magnetic nanoparticles (MNPs) into polyethylene glycol (PEG)-based
Christina Papista et al.
Kidney international, 88(2), 276-285 (2015-03-26)
IgA1 complexes containing deglycosylated IgA1, IgG autoantibodies, and a soluble form of the IgA receptor (sCD89), are hallmarks of IgA nephropathy (IgAN). Food antigens, notably gluten, are associated with increased mucosal response and IgAN onset, but their implication in the
Masaya Yokota et al.
Arthritis research & therapy, 16(2), R72-R72 (2014-03-19)
In addition to the pivotal roles of mast cells in allergic diseases, recent data suggest that mast cells play crucial roles in a variety of autoimmune responses. However, their roles in the pathogenesis of autoimmune skeletal muscle diseases have not

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service