P5115
Polypep® Low Viscosity
histochemical stabilizer
Synonym(s):
Low Viscosity Polypeptide, Polypep Solution
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
form
powder
General description
A proprietary protein digest that provides increased stablization of tissue sections in histochemical studies.
Application
Polypep® Low Viscosity has been used as a component of substrate incubation medium to act as a section stabilizer. It has also been used for enzyme cytochemistry.
Legal Information
Polypep is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 14(6), 980-987 (1999-06-03)
Bone's functional competence is established and maintained, at least partly, by mechanisms involving appropriate adaptation to mechanical loading. These appear to fail in chickens selectively bred either for maximum egg (Egg-type) or meat (Meat-type) production, which show high rates of
International journal of molecular sciences, 22(6) (2021-04-04)
The purpose of the present pilot study was to evaluate the effect of a hydrogel composed of hyaluronic acid (HA) and platelet-rich plasma (PRP) as a carrier for human mesenchymal stem cells (hMSCs) for intervertebral disc (IVD) regeneration using a
Bone, 45(2), 357-366 (2009-05-05)
Bone strength is, in part, dependent on a mechanical input that regulates the (re)modelling of skeletal elements to an appropriate size and architecture to resist fracture during habitual use. The rate of longitudinal bone growth in juveniles can also affect
Lysosomal membrane stability in mussels.
ICES Journal of Marine Science (2015)
Bivalve immunity: comparisons between the marine mussel (Mytilus edulis), the edible cockle (Cerastoderma edule) and the razor-shell (Ensis siliqua)
Fish & Shellfish Immunology, 15(3), 195-210 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service