Skip to Content
Merck
All Photos(6)

Key Documents

P4886

Sigma-Aldrich

Poly-L-glutamic acid sodium salt

greener alternative

mol wt 50,000-100,000

Synonym(s):

L-Glutamic acid homopolymer sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.25

form

powder

mol wt

50,000-100,000

greener alternative product score

old score: 46
new score: 43
Find out more about DOZN™ Scoring

greener alternative product characteristics

Designing Safer Chemicals
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

color

white to off-white

application(s)

detection

greener alternative category

storage temp.

−20°C

SMILES string

N([C@@H](CCC(=O)O)C(=O)N[C@@H](CCC(=O)O)C(=O)O)C(=O)[C@@H](N)CCC(=O)O

InChI

1S/C15H23N3O10/c16-7(1-4-10(19)20)13(25)17-8(2-5-11(21)22)14(26)18-9(15(27)28)3-6-12(23)24/h7-9H,1-6,16H2,(H,17,25)(H,18,26)(H,19,20)(H,21,22)(H,23,24)(H,27,28)/t7-,8-,9-/m0/s1

InChI key

BUZMZDDKFCSKOT-CIUDSAMLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Designing Safer Chemicals”, "Safer Solvents and Auxiliaries" and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Application


  • A real-time fluorescence assay for protease activity and inhibitor screening based on the aggregation-caused quenching of a perylene probe.: Demonstrates the utility of Poly-L-glutamic acid sodium salt in developing sensitive fluorescence assays for enzyme activity, which could streamline the development of new therapeutic inhibitors (Wang Y et al., 2018).

  • Interfacial and structural characteristics of polyelectrolyte multilayers used as cushions for supported lipid bilayers.: Explores the role of Poly-L-glutamic acid sodium salt in stabilizing lipid bilayers, crucial for bio-mimetic systems and drug delivery applications (Wlodek M et al., 2017).

  • The interaction of clozapine loaded nanocapsules with the hCMEC/D3 cells - In vitro model of blood brain barrier.: Highlights the use of Poly-L-glutamic acid sodium salt for drug encapsulation, enhancing the delivery of therapeutic agents across the blood-brain barrier (Łukasiewicz S et al., 2017).

  • A Chimeric LysK-Lysostaphin Fusion Enzyme Lysing Staphylococcus aureus Cells: a Study of Both Kinetics of Inactivation and Specifics of Interaction with Anionic Polymers.: Investigates the synergistic effects of Poly-L-glutamic acid sodium salt in enhancing the antimicrobial properties of bio-engineered enzymes, presenting a novel approach to combat bacterial infections (Filatova LY et al., 2016).

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alessandra Quarta et al.
ACS applied materials & interfaces, 9(40), 35095-35104 (2017-09-01)
In this work, the versatility of layer-by-layer technology was combined with the magnetic response of iron oxide nanobeads to prepare magnetic mesostructures with a degradable multilayer shell into which a dye quenched ovalbumin conjugate (DQ-OVA) was loaded. The system was
Marek Piotrowski et al.
Colloids and surfaces. B, Biointerfaces, 135, 8-17 (2015-08-02)
The main objectives of the present study were to investigate the biocompatibility of polyelectrolyte-coated nanocapsules and to evaluate the neuroprotective action of the nanoencapsulated water-insoluble neuroprotective drug-undecylenic acid (UDA), in vitro. Core-shell nanocapsules were synthesized using nanoemulsification and the layer-by-layer

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service