Skip to Content
Merck
All Photos(2)

Key Documents

P4405

Sigma-Aldrich

Podophyllotoxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22O8
CAS Number:
Molecular Weight:
414.41
Beilstein:
99163
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

mp

183-184 °C (lit.)

storage temp.

2-8°C

SMILES string

COc1cc(cc(OC)c1OC)[C@H]2C3C(COC3=O)[C@@H](O)c4cc5OCOc5cc24

InChI

1S/C22H22O8/c1-25-16-4-10(5-17(26-2)21(16)27-3)18-11-6-14-15(30-9-29-14)7-12(11)20(23)13-8-28-22(24)19(13)18/h4-7,13,18-20,23H,8-9H2,1-3H3/t13-,18+,19-,20-/m0/s1

InChI key

YJGVMLPVUAXIQN-XVVDYKMHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Podophyllotoxin has been used as a reference compound for evaluating the cytotoxicity in VERO cells. It has also been used as a reference standard in high-performance liquid chromatography (HPLC).

Biochem/physiol Actions

Podophyllotoxin is a naturally occurring lignan obtained from resin podophyllin present in the genus Podophyllum. It exhibits antiviral and antimitotic properties. Podophyllotoxin may be used to treat anogenital warts in children and dermatological conditions caused by psoriasis vulgaris. It may also be used as a therapeutic agent to treat genital tumors, Wilms tumors, lung cancer, and lymphomas.
Inhibits microtubule assembly; antineoplastic.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diterpenoids as potential anti-malarial compounds from Andrographis paniculata
Dwivedi M, et al.
Beni-Suef University Journal of Basic and Applied Sciences, 10(1), 1-16 (2021)
Podophyllotoxin: current perspectives
Qian L, et al.
Current Bioactive Compounds, 3(1), 37-66 (2007)
High-elevation cultivation increases anti-cancer podophyllotoxin accumulation in Podophyllum hexandrum
Li M, et al.
Industrial Crops and Products, 121, 33-344 (2018)
Ronald W Brown et al.
RSC medicinal chemistry, 11(12), 1413-1422 (2021-06-08)
African sleeping sickness is a potentially fatal neglected disease affecting sub-Saharan Africa. High-throughput screening identified the thiazolyl-benzothiophenamide 1 to be active against the causative parasite, Trypanosoma brucei. This work establishes structure-activity relationships of 1, guiding the design of second generation
Manish Kumar Dwivedi et al.
Journal of ethnopharmacology, 275, 114076-114076 (2021-04-01)
Limited drugs, rise in drug resistance against frontline anti-malarial drugs, non-availability of efficacious vaccines and high cost of drug development hinders malaria intervention programs. Search for safe, effective and affordable plant based anti-malarial agents, thus becomes crucial and vital in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service