Skip to Content
Merck
All Photos(1)

Key Documents

P3397

Sigma-Aldrich

Phosphoglucomutase from rabbit muscle

ammonium sulfate suspension, ≥100 units/mg protein

Synonym(s):

PGM, α-D-Glucose-1,6-bisphosphatase, α-D-Glucose-1-phosphate phosphotransferase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

rabbit muscle

description

ammonium sulfate suspension, >=100 units/mg protein

form

ammonium sulfate suspension

specific activity

≥100 units/mg protein

storage condition

(Tightly closed)

technique(s)

activity assay: suitable

color

white to light yellow

foreign activity

lactic dehydrogenase ≤0.5%
phosphoglucose isomerase ≤0.01%
pyruvate kinase ≤0.05%

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Research area: Cell Signaling

Phosphoglucomutase (PGM), a conserved enzyme, is abundantly found in animals, plants, and microorganisms. It is distributed in almost all tissues.

Application

Phosphoglycomutase has been used:

  • to study glycogenesis and phosphoglycomutase deficiency in humans
  • in phosphoglucomutase assays to study metabolic regulation
  • for assay A to assay enzymes closely linked to glycogen metabolism in transformed BL21(DE3)C43 cells
  • in sucrose synthase (SuSy) andADPglucose pyrophosphorylase (AGPase) assays(3)
  • in enzyme activity assay for coupling the reduction of nicotinamide adenine dinucleotide phosphate (NADP+)to determine glucose-1-phosphate from sucrose and inorganic phosphate (Pi)

Biochem/physiol Actions

Phosphoglucomutase(PGM) enzyme plays a vital role in glycolysis and gluconeogenesis. It is involved in glycogen and trehalose metabolism in insects. PGM participates in the development of plants and some microorganisms. It is essential for proteins, lipids, and nucleic acid metabolism and is crucial for the development of plants because glucose-6-phosphate is a crucial central metabolite. PGM catalyzes the interconversion of glucose-6-phosphate (G-6-P)and glucose-1-phosphate (G-1-P).

Unit Definition

One unit will convert 1.0 μmole of α-D-glucose 1-phosphate to α-D-glucose 6-phosphate per min at pH 7.4 at 30 °C.

Physical form

Crystalline suspension in 3.2 M (NH4)2SO4, pH 6.0 containing 0.01% EDTA

Analysis Note

Protein determined by biuret.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular cloning of a gene encoding the sucrose phosphorylase from Leuconostoc mesenteroides B-1149 and the expression in Escherichia coli
Lee Ha J, et al.
Enzyme and Microbial Technology, 39, 612-620 (2006)
H Sugie et al.
Neurology, 38(4), 602-605 (1988-04-01)
We report a 5-month-old boy with recurrent vomiting, lethargy, and poor weight gain. He had profound metabolic acidosis and nonketotic dicarboxylic aciduria. The serum and muscle carnitine levels were significantly low (60% and 10% of the control means, respectively), suggesting
Nora Alonso-Casajús et al.
Journal of bacteriology, 188(14), 5266-5272 (2006-07-04)
To understand the biological function of bacterial glycogen phosphorylase (GlgP), we have produced and characterized Escherichia coli cells with null or altered glgP expression. glgP deletion mutants (DeltaglgP) totally lacked glycogen phosphorylase activity, indicating that all the enzymatic activity is
Phosphoglucomutase; mechanism of action.
V JAGANNATHAN et al.
The Journal of biological chemistry, 179(2), 569-575 (1949-06-01)
Bi-Ying Pan et al.
3 Biotech, 10(2), 61-61 (2020-02-08)
Phosphoglucomutase (PGM) is a key enzyme in glycolysis and gluconeogenesis, regulating both glycogen and trehalose metabolism in insects. In this study, we explored the potential function of phosphoglucomutase (PGM) using RNA interference technology in Nilaparvata lugens, the brown planthopper. PGM1

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service