Skip to Content
Merck
All Photos(1)

Key Documents

P1411

Sigma-Aldrich

Polymyxin B-Agarose

aqueous glycerol suspension

Synonym(s):

PMB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500
NACRES:
NA.56

biological source

synthetic (organic)

Quality Level

form

aqueous glycerol suspension

extent of labeling

~1 mg per mL

technique(s)

affinity chromatography: suitable

matrix

Cross-linked 4% beaded agarose

matrix activation

cyanogen bromide

matrix attachment

amino

matrix spacer

1 atom

capacity

200-500 μg/mL binding capacity (lipopolysaccharide)(from Escherichia coli serotype 0128:B12)

suitability

suitable for chromatography

storage temp.

2-8°C

Application

Polymyxin B-agarose is used in protein chromatography, affinity chromatography and specialty resins. Polymyxin B-agarose has been used to study periodontitis and endodontic infections as well as obesity and type 2 diabetes.
Useful for removal of endotoxins from solutions

Physical form

Suspension in 50% glycerol containing 0.02% sodium azide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dina B AbuSamra et al.
Cells, 10(2) (2021-02-13)
Chemokines are an extended group of chemoattractant cytokines responsible for the recruitment of leukocytes into tissues. Among them, interferon-γ-inducible protein 10 (CXCL10) is abundantly expressed following inflammatory stimuli and participates in the trafficking of monocytes and activated T cells into
A C Issekutz
Journal of immunological methods, 61(3), 275-281 (1983-07-29)
Endotoxins liberated by gram-negative bacteria are frequent contaminants of aqueous and physiological solutions. Because of their potent biological effects in vivo and in vitro, it is often necessary to eliminate even minute quantities of endotoxin from such solutions. A process
Martín López et al.
Journal of immunology (Baltimore, Md. : 1950), 170(5), 2409-2416 (2003-02-21)
Macrophages infected with Mycobacterium tuberculosis undergo increased rates of apoptosis. Important objectives are to define the microbial factors that cause apoptosis, the mechanisms involved and the impact on infection. The 19-kDa M. tuberculosis glycolipoprotein (p19) is both cell wall-associated and
P J Jagielo et al.
The American journal of physiology, 270(6 Pt 1), L1052-L1059 (1996-06-01)
To investigate the role of endotoxin in grain dust-induced airway inflammation, we reduced the endotoxin activity from extracts of corn dust (CDE), using three distinct methods, and determined the effect of endotoxin activity on the in vitro and in vivo
K W Talmadge et al.
Journal of chromatography, 476, 175-185 (1989-08-04)
A new affinity column packing for removal of endotoxins has been prepared by coupling USP drug-quality polymyxin B to Affi-Prep, a novel synthetic macroporous polymer. Affi-Prep Polymyxin binds endotoxins from a number of different strains of gram-negative bacteria. Endotoxin binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service