Skip to Content
Merck
All Photos(1)

Documents

P0928

Sigma-Aldrich

Resorufin pentyl ether

Synonym(s):

7-Pentyloxy-3-phenoxazone, 7-Pentyloxy-3H-phenoxazin-3-one, O7-Pentylresorufin, Pentoxyresorufin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17NO3
CAS Number:
Molecular Weight:
283.32
Beilstein:
8394939
MDL number:
UNSPSC Code:
12161501
PubChem Substance ID:
NACRES:
NA.47

Assay

≥98% (TLC)

Quality Level

form

powder

solubility

acetonitrile: 0.95- 1.05 mg/mL, clear, orange

storage temp.

2-8°C

SMILES string

CCCCCOc1ccc2N=C3C=CC(=O)C=C3Oc2c1

InChI

1S/C17H17NO3/c1-2-3-4-9-20-13-6-8-15-17(11-13)21-16-10-12(19)5-7-14(16)18-15/h5-8,10-11H,2-4,9H2,1H3

InChI key

ZPSOKQFFOYYPKC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Resorufin pentyl ether is an analog of resazurin. Resorufin is a redox probe which is colorless and non-fluorescent in its reduced state. On oxidation, it fluoresces red. Resorufin is used widely as an indicator in microbiological media. It can be used to differentiate between actively metabolizing cells and inactive or dead cells.

Application

Resorufin pentyl ether has been used as a substrate to study the effects of Kaempferia parviflora extract on mouse hepatic cytochrome P450 enzymes.

Biochem/physiol Actions

Fluorimetric substrate for cytochrome P450 linked enzymes, CYP2B and CYP2B4.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Advances in Microbial Ecology null
E S Tzanakakis et al.
Cell motility and the cytoskeleton, 48(3), 175-189 (2001-02-27)
Cultured rat hepatocytes self-assemble into three-dimensional structures or spheroids that exhibit ultrastructural characteristics of native hepatic tissue and enhanced liver-specific functions. The spheroid formation process involves cell translocation and changes in cell shape, indicative of the reorganization of the cytoskeletal
Microbial Biofilms null
R B van Breemen et al.
Drug metabolism and disposition: the biological fate of chemicals, 26(2), 85-90 (1998-03-28)
An on-line mass spectrometric method has been developed to generate and identify drug metabolites formed by hepatic cytochromes P450. This method, pulsed ultrafiltration-mass spectrometry, may be used for rapid screening of drugs to determine their extent of metabolism by microsomal
Canan Sapmaz et al.
Drug and chemical toxicology, 43(1), 13-21 (2018-05-19)
Morin is a flavonoid which is present in many plants. Endosulfan and 7,12-dimethylbenz[a]anthracene (DMBA) are toxic chemicals that humans are exposed to in their daily lives. In this study, the protective role of morin was investigated in endosulfan and DMBA

Articles

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Phase I biotransformation reactions increase drug compound polarity, mainly occurring in hepatic circulation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service